Learn More
Thermo Scientific™ Hypersil GOLD™ Amino HPLC Columns
Achieve outstanding peak shape and sensitivity in weak anion exchange, reversed phase and normal phase modes with Hypersil GOLD Amino LC Columns.
£268.06 - £958.65
Specifications
pH | 2 to 8 |
---|---|
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
USP Type | L8 |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10147654
|
Thermo Scientific™
25705-104630 |
5 μm | 4.6 mm | 100 mm |
£810.57
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
10006074
|
Thermo Scientific™
25705-154630 |
5 μm | 4.6 mm | 150 mm |
£676.30
Each |
In Stock
Log in to see stock. |
|||
10322975
|
Thermo Scientific™
25705-254030 |
5 μm | 4 mm | 250 mm |
£712.55
Each |
In Stock
Log in to see stock. |
|||
10752717
|
Thermo Scientific™
25705-254630 |
5 μm | 4.6 mm | 250 mm |
£712.55
Each |
In Stock
Log in to see stock. |
|||
10732347
|
Thermo Scientific™
25702-202130 |
1.9 μm | 2.1 mm | 200 mm |
£958.65
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
10670246
|
Thermo Scientific™
25702-152130 |
1.9 μm | 2.1 mm | 150 mm |
£915.73
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10177994
|
Thermo Scientific™
25702-102130 |
1.9 μm | 2.1 mm | 100 mm |
£825.11
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10793097
|
Thermo Scientific™
25702-052130 |
1.9 μm | 2.1 mm | 50 mm |
£833.69
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
10381295
|
Thermo Scientific™
25703-154630 |
3 μm | 4.6 mm | 150 mm |
£727.82
Each |
In Stock
Log in to see stock. |
|||
10169564
|
Thermo Scientific™
25703-153030 |
3 μm | 3 mm | 150 mm |
£779.33
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10600826
|
Thermo Scientific™
25703-152130 |
3 μm | 2.1 mm | 150 mm |
£779.33
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
10289364
|
Thermo Scientific™
25703-104630 |
3 μm | 4.6 mm | 100 mm |
£810.57
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10056114
|
Thermo Scientific™
25703-014001 |
3 μm | 4 mm | 10 mm |
£395.81
Pack of 4 |
In Stock
Log in to see stock. |
|||
10478664
|
Thermo Scientific™
25703-013001 |
3 μm | 3 mm | 10 mm |
£392.05
Pack of 4 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10259174
|
Thermo Scientific™
25705-014001 |
5 μm | 4 mm | 10 mm |
£356.76
Pack of 4 |
In Stock
Log in to see stock. |
|||
10517455
|
Thermo Scientific™
25705-253030 |
5 μm | 3 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10742717
|
Thermo Scientific™
25705-152130 |
5 μm | 2.1 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10211095
|
Thermo Scientific™
25703-151030 |
3 μm | 1 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10157554
|
Thermo Scientific™
25705-054630 |
5 μm | 4.6 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10689755
|
Thermo Scientific™
25703-103030 |
3 μm | 3 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10742527
|
Thermo Scientific™
25703-102130 |
3 μm | 2.1 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10187654
|
Thermo Scientific™
25703-052130 |
3 μm | 2.1 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10681196
|
Thermo Scientific™
25703-032130 |
3 μm | 2.1 mm | 30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10554105
|
Thermo Scientific™
25705-252130 |
5 μm | 2.1 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10391295
|
Thermo Scientific™
25705-102130 |
5 μm | 2.1 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10698515
|
Thermo Scientific™
25703-012101 |
3 μm | 2.1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10036304
|
Thermo Scientific™
25703-011001 |
3 μm | 1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10427324
|
Thermo Scientific™
25705-052130 |
5 μm | 2.1 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10525615
|
Thermo Scientific™
25705-013001 |
5 μm | 3 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10659465
|
Thermo Scientific™
25705-012101 |
5 μm | 2.1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Achieve excellent chromatographic separations in weak anion exchange, reversed phase and normal phase modes with Thermo Scientific™ Hypersil GOLD™ Amino LC Columns. These columns, featuring a high-performance aminopropyl phase, provide outstanding peak shape and sensitivity and are available with a 1.9μm particle size for high speed, high efficiency separations.
Excellent Chromatographic Properties in Three Modes- Retains anions and organic acids in weak anion exchange mode when used with common buffers and an organic modifier
- Excels for carbohydrate analysis in reversed phase mode
- Offers an alternative selectivity to silica columns in normal phase chromatography
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
- Excellent peak symmetry
- Narrow peaks for outstanding efficiency
- Enhanced sensitivity and improved resolution
- Exceptional reproducibility for reliable chromatography, column after column
- Solutions for high-throughput screening
- Providing 1.9, 3, and 5μm particle sizes for all your separation needs
Specifications
2 to 8 | |
220 m²/g | |
L8 | |
2% | |
Yes |
175 Å | |
60°C | |
Amino (NH2) | |
Mixed Mode | |
Reversed Phase, Normal Phase, Ion Exchange, HILIC |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.