Learn More
Thermo Scientific™ Hypersil GOLD™ aQ C18 Polar Endcapped HPLC Columns
Retain and resolve polar analytes with highly aqueous mobile phases.
£356.76 - £12555.00
Specifications
pH | 2 to 9 |
---|---|
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
USP Type | L1 |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10783497
|
Thermo Scientific™
25305-012101 |
5 μm | 2.1 mm | 10 mm |
£356.76
Pack of 4 |
In Stock
Log in to see stock. |
|||
10582065
|
Thermo Scientific™
25305-254630 |
5 μm | 4.6 mm | 250 mm |
£698.72
Each |
In Stock
Log in to see stock. |
|||
10161023
|
Thermo Scientific™
25305-252130 |
5 μm | 2.1 mm | 250 mm |
£751.66
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10019382
|
Thermo Scientific™
25305-154630 |
5 μm | 4.6 mm | 150 mm |
£676.30
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
10497414
|
Thermo Scientific™
25305-153030 |
5 μm | 3 mm | 150 mm |
£709.69
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
10718063
|
Thermo Scientific™
25305-152130 |
5 μm | 2.1 mm | 150 mm |
£709.69
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10150593
|
Thermo Scientific™
25305-104630 |
5 μm | 4.6 mm | 100 mm |
£640.05
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10557435
|
Thermo Scientific™
25305-103030 |
5 μm | 3 mm | 100 mm |
£724.02
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10140593
|
Thermo Scientific™
25305-102130 |
5 μm | 2.1 mm | 100 mm |
£687.56
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10662474
|
Thermo Scientific™
25305-054630 |
5 μm | 4.6 mm | 50 mm |
£587.59
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10508405
|
Thermo Scientific™
25305-053030 |
5 μm | 3 mm | 50 mm |
£662.60
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10776153
|
Thermo Scientific™
25305-052130 |
5 μm | 2.1 mm | 50 mm |
£617.16
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
11578280
|
Thermo Scientific™
25305-014001 |
5 μm | 4 mm | 10 mm |
£356.76
Pack of 4 |
In Stock
Log in to see stock. |
|||
10370905
|
Thermo Scientific™
25305-013001 |
5 μm | 3 mm | 10 mm |
£380.83
Pack of 4 |
In Stock
Log in to see stock. |
|||
10008902
|
Thermo Scientific™
25303-154630 |
3 μm | 4.6 mm | 150 mm |
£742.12
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10509355
|
Thermo Scientific™
25303-154030 |
3 μm | 4 mm | 150 mm |
£846.87
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
10268944
|
Thermo Scientific™
25303-153030 |
3 μm | 3 mm | 150 mm |
£779.33
Each |
In Stock
Log in to see stock. |
|||
10572255
|
Thermo Scientific™
25303-152130 |
3 μm | 2.1 mm | 150 mm |
£779.33
Each |
In Stock
Log in to see stock. |
|||
10210655
|
Thermo Scientific™
25303-151030 |
3 μm | 1 mm | 150 mm |
£950.16
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10364603
|
Thermo Scientific™
25303-104630 |
3 μm | 4.6 mm | 100 mm |
£699.20
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
10139514
|
Thermo Scientific™
25303-104030 |
3 μm | 4 mm | 100 mm |
£860.82
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10074313
|
Thermo Scientific™
25303-103030 |
3 μm | 3 mm | 100 mm |
£738.31
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
10190873
|
Thermo Scientific™
25303-102130 |
3 μm | 2.1 mm | 100 mm |
£738.31
Each |
In Stock
Log in to see stock. |
|||
10541105
|
Thermo Scientific™
25303-054630 |
3 μm | 4.6 mm | 50 mm |
£735.19
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10629955
|
Thermo Scientific™
25303-054030 |
3 μm | 4 mm | 50 mm |
£802.19
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
10251235
|
Thermo Scientific™
25303-053030 |
3 μm | 3 mm | 50 mm |
£735.19
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
10513795
|
Thermo Scientific™
25303-052130 |
3 μm | 2.1 mm | 50 mm |
£678.21
Each |
In Stock
Log in to see stock. |
|||
10715793
|
Thermo Scientific™
25303-032130 |
3 μm | 2.1 mm | 30 mm |
£678.21
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
11568280
|
Thermo Scientific™
25303-014001 |
3 μm | 4 mm | 10 mm |
£392.05
Pack of 4 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
11558280
|
Thermo Scientific™
25303-013001 |
3 μm | 3 mm | 10 mm |
£392.05
Pack of 4 |
In Stock
Log in to see stock. |
|||
11548280
|
Thermo Scientific™
25303-012101 |
3 μm | 2.1 mm | 10 mm |
£392.05
Pack of 4 |
In Stock
Log in to see stock. |
|||
10360905
|
Thermo Scientific™
25303-011001 |
3 μm | 1 mm | 10 mm |
£387.88
Pack of 4 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
10486693
|
Thermo Scientific™
25302-202130 |
1.9 μm | 2.1 mm | 200 mm |
£919.55
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
10736734
|
Thermo Scientific™
25302-152130 |
1.9 μm | 2.1 mm | 150 mm |
£915.73
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10631344
|
Thermo Scientific™
25302-103030 |
1.9 μm | 3 mm | 100 mm |
£833.69
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
10582635
|
Thermo Scientific™
25302-102130 |
1.9 μm | 2.1 mm | 100 mm |
£825.11
Each |
In Stock
Log in to see stock. |
|||
10621344
|
Thermo Scientific™
25302-101030 |
1.9 μm | 1 mm | 100 mm |
£833.69
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10119574
|
Thermo Scientific™
25302-054630 |
1.9 μm | 4.6 mm | 50 mm |
£725.91
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
10111723
|
Thermo Scientific™
25302-053030 |
1.9 μm | 3 mm | 50 mm |
£905.49
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10572635
|
Thermo Scientific™
25302-052130 |
1.9 μm | 2.1 mm | 50 mm |
£817.51
Each |
In Stock
Log in to see stock. |
|||
10101723
|
Thermo Scientific™
25302-051030 |
1.9 μm | 1 mm | 50 mm |
£835.60
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10324223
|
Thermo Scientific™
25302-032130 |
1.9 μm | 2.1 mm | 30 mm |
£833.94
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
10129514
|
Thermo Scientific™
25303-101030 |
3 μm | 1 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10362525
|
Thermo Scientific™
25305-109570 |
5 μm | 50 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10109374
|
Thermo Scientific™
25305-159070 |
5 μm | 10 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10221325
|
Thermo Scientific™
25305-159270 |
5 μm | 21.2 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10017824
|
Thermo Scientific™
25305-159370 |
5 μm | 30 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10027824
|
Thermo Scientific™
25305-159570 |
5 μm | 50 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10231085
|
Thermo Scientific™
25305-259070 |
5 μm | 10 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10241085
|
Thermo Scientific™
25305-259270 |
5 μm | 21.2 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10117664
|
Thermo Scientific™
25305-019023 |
5 μm | 10 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10527245
|
Thermo Scientific™
25305-259370 |
5 μm | 30 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10446844
|
Thermo Scientific™
25305-259570 |
5 μm | 50 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10392955
|
Thermo Scientific™
25305-059070 |
5 μm | 10 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10659945
|
Thermo Scientific™
25305-059270 |
5 μm | 21.2 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10017874
|
Thermo Scientific™
25305-059370 |
5 μm | 30 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10056134
|
Thermo Scientific™
25305-254030 |
5 μm | 4 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10611296
|
Thermo Scientific™
25305-253030 |
5 μm | 3 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10538395
|
Thermo Scientific™
25305-059570 |
5 μm | 50 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10649215
|
Thermo Scientific™
25305-109270 |
5 μm | 21.2 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10363105
|
Thermo Scientific™
25305-154030 |
5 μm | 4 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10448564
|
Thermo Scientific™
25305-109370 |
5 μm | 30 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10375203
|
Thermo Scientific™
25302-022130 |
1.9 μm | 2.1 mm | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Beschreibung
Retain and resolve polar analytes using Thermo Scientific™ Hypersil GOLD™ aQ Polar Endcapped C18 columns. These columns provide excellent peak shape, even with challenging reversed phase separations using highly aqueous mobile phases.
Alternative selectivity with polar endcapped C18 stationary phase- Polar functional group in endcap provides additional interaction mechanism for separation of polar compounds
- Yields superior retention of polar compounds
- Usable in 100% aqueous mobile phases
- No loss of performance or stability
- Controlled secondary interactions ensure optimal peak shape for all analyte types
- Ideal for the quantitative analysis of trace levels of polar analytes
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
- Excellent peak symmetry
- Narrow peaks for outstanding efficiency
- Enhanced sensitivity and improved resolution
- Exceptional reproducibility for reliable chromatography, column after column
- Solutions for high-throughput screening
- 1.9, 3, and 5 μm particle sizes for all your separation needs
Spezifikation
2 to 9 | |
220 m²/g | |
L1 | |
Reversed Phase | |
Reversed Phase |
175 Å | |
60°C | |
12% | |
Polar |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.