Learn More
Thermo Scientific™ Hypersil GOLD™ C4 HPLC Columns
Featuring a lower hydrophobicity than C18 or C8 columns, Thermo Scientific™ Hypersil GOLD™ C4 columns provide outstanding sensitivity and high efficiency.
Brand: Thermo Scientific™ 25502-102130
Code : 16
Additional Details : Weight : 0.45360kg
Description
For very hydrophobic analytes, achieve faster separations with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns. Featuring a lower hydrophobicity than C18 or C8 columns, Hypersil GOLD C4 columns provide outstanding sensitivity and high efficiency.
Similar selectivity, less retention than C18 or C8 columns- Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
- Ideal for analysis of hydrophobic polypeptides and proteins
- Highly symmetrical peak shapes
- 1.9μm particle size columns improve speed and efficiency
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
- Excellent peak symmetry
- Narrow peaks for outstanding efficiency
- Enhanced sensitivity and improved resolution
- Exceptional reproducibility for reliable chromatography, column after column
- Solutions for high-throughput screening
- 1.9, 3, and 5μm particle sizes for all your separation needs
Specifications
Analytical Column | |
0.55 mL/min | |
100 mm | |
1.9 μm | |
175 Å | |
60°C | |
C4 | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
2.1 mm | |
HPLC | |
18130 psi (1250 bar) | |
2 to 8 | |
220 m²/g | |
L26 | |
Hypersil GOLD | |
5% | |
Yes |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.