Learn More
Thermo Scientific™ Hypersil GOLD™ C4 HPLC Columns
Featuring a lower hydrophobicity than C18 or C8 columns, Thermo Scientific™ Hypersil GOLD™ C4 columns provide outstanding sensitivity and high efficiency.
£271.32 - £934.65
Specifications
pH | 2 to 8 |
---|---|
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
USP Type | L26 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||||
10609585
|
Thermo Scientific™
25505-052130 |
50 mm | 2.1 mm | 5 μm | N/A | ||||||
10699385
|
Thermo Scientific™
25505-054630 |
50 mm | 4.6 mm | 5 μm | N/A | ||||||
10382865
|
Thermo Scientific™
25505-102130 |
100 mm | 2.1 mm | 5 μm | N/A | ||||||
10322355
|
Thermo Scientific™
25505-103030 |
100 mm | 3 mm | 5 μm | N/A | ||||||
10699755
|
Thermo Scientific™
25505-104630 |
100 mm | 4.6 mm | 5 μm | N/A | ||||||
10609765
|
Thermo Scientific™
25505-152130 |
150 mm | 2.1 mm | 5 μm | N/A | ||||||
10688435
|
Thermo Scientific™
25505-154030 |
150 mm | 4 mm | 5 μm | N/A | ||||||
10486854
|
Thermo Scientific™
25505-154630 |
150 mm | 4.6 mm | 5 μm |
£648.23
Each |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
10067684
|
Thermo Scientific™
25505-252130 |
250 mm | 2.1 mm | 5 μm | N/A | ||||||
10312635
|
Thermo Scientific™
25505-254630 |
250 mm | 4.6 mm | 5 μm |
£683.40
Each |
In Stock
Log in to see stock. |
|||||
10127614
|
Thermo Scientific™
25502-052130 |
50 mm | 2.1 mm | 1.9 μm |
£860.27
Each |
In Stock
Log in to see stock. |
|||||
10691396
|
Thermo Scientific™
25502-102130 |
100 mm | 2.1 mm | 1.9 μm |
£849.23
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
10391585
|
Thermo Scientific™
25502-152130 |
150 mm | 2.1 mm | 1.9 μm |
£934.65
Each |
In Stock
Log in to see stock. |
|||||
10147884
|
Thermo Scientific™
25503-011001 |
10 mm | 1 mm | 3 μm | N/A | ||||||
10447534
|
Thermo Scientific™
25503-012101 |
10 mm | 2.1 mm | 3 μm |
£374.87
Pack of 4 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
10157884
|
Thermo Scientific™
25503-013001 |
10 mm | 3 mm | 3 μm |
£374.87
Pack of 4 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
10610626
|
Thermo Scientific™
25503-014001 |
10 mm | 4 mm | 3 μm | N/A | ||||||
10332835
|
Thermo Scientific™
25503-032130 |
30 mm | 2.1 mm | 3 μm | N/A | ||||||
10159534
|
Thermo Scientific™
25503-052130 |
50 mm | 2.1 mm | 3 μm |
£648.23
Each |
In Stock
Log in to see stock. |
|||||
10037254
|
Thermo Scientific™
25503-102130 |
100 mm | 2.1 mm | 3 μm | N/A | ||||||
10027504
|
Thermo Scientific™
25503-103030 |
100 mm | 3 mm | 3 μm | N/A | ||||||
10732826
|
Thermo Scientific™
25503-104630 |
100 mm | 4.6 mm | 3 μm | N/A | ||||||
10199474
|
Thermo Scientific™
25503-151030 |
150 mm | 1 mm | 3 μm | N/A | ||||||
10599735
|
Thermo Scientific™
25503-152130 |
150 mm | 2.1 mm | 3 μm | N/A | ||||||
10312355
|
Thermo Scientific™
25503-153030 |
150 mm | 3 mm | 3 μm | N/A | ||||||
10261325
|
Thermo Scientific™
25503-154630 |
150 mm | 4.6 mm | 3 μm |
£713.55
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
10076694
|
Thermo Scientific™
25505-012101 |
10 mm | 2.1 mm | 5 μm | N/A | ||||||
10670046
|
Thermo Scientific™
25505-014001 |
10 mm | 4 mm | 5 μm | N/A | ||||||
Description
For very hydrophobic analytes, achieve faster separations with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns. Featuring a lower hydrophobicity than C18 or C8 columns, Hypersil GOLD C4 columns provide outstanding sensitivity and high efficiency.
Similar selectivity, less retention than C18 or C8 columns- Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
- Ideal for analysis of hydrophobic polypeptides and proteins
- Highly symmetrical peak shapes
- 1.9 μm particle size columns improve speed and efficiency
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
- Excellent peak symmetry
- Narrow peaks for outstanding efficiency
- Enhanced sensitivity and improved resolution
- Exceptional reproducibility for reliable chromatography, column after column
- Solutions for high-throughput screening
- 1.9, 3, and 5 μm particle sizes for all your separation needs
Specifications
2 to 8 | |
220 m²/g | |
L26 | |
5% | |
Yes |
175 Å | |
60°C | |
C4 | |
Reversed Phase | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.