Learn More
Thermo Scientific™ Hypersil GOLD™ C8 HPLC Columns
Have analytes with medium hydrophobicity or need a less hydrophobic phase to obtain optimum retention? These columns provide outstanding sensitivity and excellent peak shape.
£356.76 - £1270.00
Specifications
pH | 2 to 9 |
---|---|
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
USP Type | L7 |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Packing Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Packing Material | Price | Quantity & Availability | ||
10503235
|
Thermo Scientific™
25205-102130 |
5 μm | 2.1 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
£674.40
Each |
In Stock
Log in to see stock. |
||
10343465
|
Thermo Scientific™
25205-254030 |
5 μm | 4 mm | 250 mm | Spherical, Fully Porous, Ultrapure Silica |
£712.55
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10755783
|
Thermo Scientific™
25205-154630 |
5 μm | 4.6 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
£676.30
Each |
In Stock
Log in to see stock. |
||
10690564
|
Thermo Scientific™
25205-054630 |
5 μm | 4.6 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
£587.59
Each |
In Stock
Log in to see stock. |
||
11538380
|
Thermo Scientific™
25205-022130 |
5 μm | 2.1 mm | 20 mm | Silica |
£617.16
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
10293413
|
Thermo Scientific™
25205-052130 |
5 μm | 2.1 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
£617.16
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10203663
|
Thermo Scientific™
25205-152130 |
5 μm | 2.1 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
£709.69
Each |
In Stock
Log in to see stock. |
||
10243453
|
Thermo Scientific™
25205-254630 |
5 μm | 4.6 mm | 250 mm | Spherical, Fully Porous, Ultrapure Silica |
£712.55
Each |
In Stock
Log in to see stock. |
||
10690754
|
Thermo Scientific™
25205-104630 |
5 μm | 4.6 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
£640.05
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
10736144
|
Thermo Scientific™
25203-154630 |
3 μm | 4.6 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
£742.12
Each |
In Stock
Log in to see stock. |
||
10312745
|
Thermo Scientific™
25203-153030 |
3 μm | 3 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
£779.33
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10078952
|
Thermo Scientific™
25203-152130 |
3 μm | 2.1 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
£779.33
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10352695
|
Thermo Scientific™
25203-151030 |
3 μm | 1 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
£950.16
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10774414
|
Thermo Scientific™
25203-104630 |
3 μm | 4.6 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
£705.88
Each |
In Stock
Log in to see stock. |
||
10619395
|
Thermo Scientific™
25203-103030 |
3 μm | 3 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
£738.31
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
10018912
|
Thermo Scientific™
25203-102130 |
3 μm | 2.1 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
£738.31
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10543995
|
Thermo Scientific™
25203-054630 |
3 μm | 4.6 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
£645.78
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10558585
|
Thermo Scientific™
25203-053030 |
3 μm | 3 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
£678.21
Each |
In Stock
Log in to see stock. |
||
10553035
|
Thermo Scientific™
25203-052130 |
3 μm | 2.1 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
£671.53
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10283653
|
Thermo Scientific™
25203-034630 |
3 μm | 4.6 mm | 30 mm | Spherical, Fully Porous, Ultrapure Silica |
£699.83
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10119564
|
Thermo Scientific™
25203-033030 |
3 μm | 3 mm | 30 mm | Spherical, Fully Porous, Ultrapure Silica |
£727.74
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10234864
|
Thermo Scientific™
25202-152130 |
1.9 μm | 2.1 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
£915.73
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10074694
|
Thermo Scientific™
25202-102130 |
1.9 μm | 2.1 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
£825.11
Each |
In Stock
Log in to see stock. |
||
10329884
|
Thermo Scientific™
25202-101030 |
1.9 μm | 1 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
£1004.14
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10664945
|
Thermo Scientific™
25202-052130 |
1.9 μm | 2.1 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
£833.69
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
11508280
|
Thermo Scientific™
25205-014001 |
5 μm | 4 mm | 10 mm | Silica |
£356.76
Pack of 4 |
In Stock
Log in to see stock. |
||
10259644
|
Thermo Scientific™
25205-013001 |
5 μm | 3 mm | 10 mm | High Purity Silica |
£380.83
Pack of 4 |
In Stock
Log in to see stock. |
||
10535445
|
Thermo Scientific™
25205-012101 |
5 μm | 2.1 mm | 10 mm | Silica |
£356.76
Pack of 4 |
In Stock
Log in to see stock. |
||
10609855
|
Thermo Scientific™
25205-011001 |
5 μm | 1 mm | 10 mm | Silica |
£356.76
Pack of 4 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10487694
|
Thermo Scientific™
25203-014001 |
3 μm | 4 mm | 10 mm | Silica |
£410.27
Pack of 4 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10075814
|
Thermo Scientific™
25203-013001 |
3 μm | 3 mm | 10 mm | Silica |
£392.05
Pack of 4 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
11598270
|
Thermo Scientific™
25203-012101 |
3 μm | 2.1 mm | 10 mm | Silica |
£392.05
Pack of 4 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
10714817
|
Thermo Scientific™
25203-011001 |
3 μm | 1 mm | 10 mm | Silica |
£418.27
Pack of 4 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
11542821
|
Thermo Scientific™
25203-254630 |
3 μm | 4.6 mm | 250 mm | Silica |
£783.14
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10442195
|
Thermo Scientific™
25202-103030 |
1.9 μm | 3 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10764356
|
Thermo Scientific™
25205-053030 |
5 μm | 3 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10202105
|
Thermo Scientific™
25203-104030 |
3 μm | 4 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10096024
|
Thermo Scientific™
25202-054630 |
1.9 μm | 4.6 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10619405
|
Thermo Scientific™
25205-253030 |
5 μm | 3 mm | 250 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10442005
|
Thermo Scientific™
25202-051030 |
1.9 μm | 1 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10044264
|
Thermo Scientific™
25202-032130 |
1.9 μm | 2.1 mm | 30 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10639565
|
Thermo Scientific™
25205-051030 |
5 μm | 1 mm | 50 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10027464
|
Thermo Scientific™
25203-101030 |
3 μm | 1 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10149494
|
Thermo Scientific™
25205-154030 |
5 μm | 4 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11543141
|
Thermo Scientific™
25205-259070 |
5 μm | 10 mm | 250 mm | Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10734766
|
Thermo Scientific™
25203-154030 |
3 μm | 4 mm | 150 mm | Silica, Spherical Fully Porous Ultrapure |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10108874
|
Thermo Scientific™
25205-153030 |
5 μm | 3 mm | 150 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10261915
|
Thermo Scientific™
25205-103030 |
5 μm | 3 mm | 100 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10746334
|
Thermo Scientific™
25203-032130 |
3 μm | 2.1 mm | 30 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10765783
|
Thermo Scientific™
25205-252130 |
5 μm | 2.1 mm | 250 mm | Spherical, Fully Porous, Ultrapure Silica |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
For analytes with medium hydrophobicity or when a less hydrophobic phase is required to obtain optimum retention, use Thermo Scientific™ Hypersil GOLD™ C8 LC columns for outstanding sensitivity and excellent peak shape. The selectivity is similar to C18 columns with a predictable elution order but less retention. The lower hydrophobicity allows compounds to elute more quickly, yielding faster separations.
Similar selectivity, less retention than C18 columns- Successful retention and separation of moderately hydrophobic analytes
- Ideal for fatty acids, which are too hydrophobic for analysis using C18 columns
- Higher analytical throughput compared to C18 alkyl chain chemistry
- Quicker compound elution due to reduced hydrophobic interactions
- Highly symmetrical peak shapes
- Improved speed of analysis, efficiency, and sensitivity
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
- Excellent peak symmetry
- Narrow peaks for outstanding efficiency
- Enhanced sensitivity and improved resolution
- Exceptional reproducibility for reliable chromatography, column after column
- Solutions for high-throughput screening
- 1.9, 3 and 5μm particle sizes for all your separation needs
Specifications
2 to 9 | |
220 m²/g | |
L7 | |
8% |
175 Å | |
60°C | |
C8 | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.