Learn More
Thermo Scientific™ Hypersil GOLD™ PEI HILIC HPLC Columns
Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Hypersil GOLD PEI HILIC LC Columns.
£268.06 - £915.73
Specifications
pH | 2 to 8 |
---|---|
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
Stationary Phase | Polyethyleneimine |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
11537971
|
Thermo Scientific™
26505-052130 |
5 μm | 2.1 mm | 50 mm |
£646.73
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11587971
|
Thermo Scientific™
26505-102130 |
5 μm | 2.1 mm | 100 mm |
£674.40
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11537981
|
Thermo Scientific™
26505-152130 |
5 μm | 2.1 mm | 150 mm |
£709.69
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11567981
|
Thermo Scientific™
26505-154630 |
5 μm | 4.6 mm | 150 mm |
£676.30
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11517991
|
Thermo Scientific™
26505-254630 |
5 μm | 4.6 mm | 250 mm |
£712.55
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10029259
|
Thermo Scientific™
26502-152130 |
1.9 μm | 2.1 mm | 150 mm |
£915.73
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11567931
|
Thermo Scientific™
26502-102130 |
1.9 μm | 2.1 mm | 100 mm |
£833.69
Each |
In Stock
Log in to see stock. |
|||
11527931
|
Thermo Scientific™
26502-052130 |
1.9 μm | 2.1 mm | 50 mm |
£833.69
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||
11527961
|
Thermo Scientific™
26503-154630 |
3 μm | 4.6 mm | 150 mm |
£742.12
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11507961
|
Thermo Scientific™
26503-153030 |
3 μm | 3 mm | 150 mm |
£794.52
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11597951
|
Thermo Scientific™
26503-152130 |
3 μm | 2.1 mm | 150 mm |
£779.33
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11587951
|
Thermo Scientific™
26503-151030 |
3 μm | 1 mm | 150 mm |
£856.58
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11557951
|
Thermo Scientific™
26503-103030 |
3 μm | 3 mm | 100 mm |
£738.31
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
11547951
|
Thermo Scientific™
26503-102130 |
3 μm | 2.1 mm | 100 mm |
£737.35
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
11597941
|
Thermo Scientific™
26503-052130 |
3 μm | 2.1 mm | 50 mm |
£678.21
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11597971
|
Thermo Scientific™
26505-103030 |
5 μm | 3 mm | 100 mm |
£674.40
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||
11748056
|
Thermo Scientific™
26503-012101 |
3 μm | 2.1 mm | 10 mm |
£392.05
Pack of 4 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11738056
|
Thermo Scientific™
26503-011001 |
3 μm | 1 mm | 10 mm |
£418.17
Pack of 4 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
11728056
|
Thermo Scientific™
26505-014001 |
5 μm | 4 mm | 10 mm |
£381.21
Pack of 4 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11507991
|
Thermo Scientific™
26505-254030 |
5 μm | 4 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11547941
|
Thermo Scientific™
26503-032130 |
3 μm | 2.1 mm | 30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11517981
|
Thermo Scientific™
26505-104630 |
5 μm | 4.6 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11567971
|
Thermo Scientific™
26505-054630 |
5 μm | 4.6 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11768056
|
Thermo Scientific™
26503-014001 |
3 μm | 4 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11758056
|
Thermo Scientific™
26503-013001 |
3 μm | 3 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11587981
|
Thermo Scientific™
26505-252130 |
5 μm | 2.1 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11577951
|
Thermo Scientific™
26503-104630 |
3 μm | 4.6 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11597981
|
Thermo Scientific™
26505-253030 |
5 μm | 3 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11708056
|
Thermo Scientific™
26505-012101 |
5 μm | 2.1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Thermo Scientific™ Hypersil GOLD™ PEI HILIC (hydrophilic interaction liquid chromatography) LC Columns. These columns deliver straightforward separation without the need for sample derivatization or ion-pair reagents. Featuring outstanding peak shape and selectivity, the columns provide improved sensitivity for MS detection. With 1.9, 3 and 5μm particle sizes, Hypersil GOLD PEI HILIC Columns are ideal for applications within drug discovery, food testing and environmental analysis.
About the Hypersil GOLD Family of HPLC Columns:Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
- Excellent peak symmetry
- Narrow peaks for outstanding efficiency
- Enhanced sensitivity and improved resolution
- Exceptional reproducibility for reliable chromatography, column after column
- Solutions for high-throughput screening
- 1.9, 3 and 5μm particle sizes for all your separation needs
Specifications
2 to 8 | |
220 m²/g | |
Polyethyleneimine | |
HILIC | |
HILIC |
175 Å | |
60°C | |
6% | |
No |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.