Learn More
Thermo Scientific™ Hypersil GOLD™ C18 Selectivity HPLC Columns
Use these endcapped, ultrapure, silica-based columns to generate exceptional peak shape and resolution for HPLC and LC/MS analyses using generic gradients
£230.66 - £12555.00
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
10211625
|
Thermo Scientific™
25003-031030 |
30 mm | 1 mm | 3 μm | N/A | ||||
10394463
|
Thermo Scientific™
25003-032130 |
30 mm | 2.1 mm | 3 μm |
£671.53
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
10508995
|
Thermo Scientific™
25003-033030 |
30 mm | 3 mm | 3 μm |
£743.59
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10764984
|
Thermo Scientific™
25003-034630 |
30 mm | 4.6 mm | 3 μm |
£645.78
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
10342595
|
Thermo Scientific™
25003-051030 |
50 mm | 1 mm | 3 μm |
£732.68
Each |
In Stock
Log in to see stock. |
|||
10424413
|
Thermo Scientific™
25003-052130 |
50 mm | 2.1 mm | 3 μm |
£678.21
Each |
In Stock
Log in to see stock. |
|||
10293183
|
Thermo Scientific™
25003-053030 |
50 mm | 3 mm | 3 μm |
£678.21
Each |
In Stock
Log in to see stock. |
|||
10658825
|
Thermo Scientific™
25003-054030 |
50 mm | 4 mm | 3 μm |
£820.39
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10089282
|
Thermo Scientific™
25003-054630 |
50 mm | 4.6 mm | 3 μm |
£645.78
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||
10281225
|
Thermo Scientific™
25003-101030 |
100 mm | 1 mm | 3 μm |
£786.26
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10632864
|
Thermo Scientific™
25003-102130 |
100 mm | 2.1 mm | 3 μm |
£738.31
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10304923
|
Thermo Scientific™
25003-103030 |
100 mm | 3 mm | 3 μm |
£738.31
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10794546
|
Thermo Scientific™
25003-104030 |
100 mm | 4 mm | 3 μm |
£705.88
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||
10384323
|
Thermo Scientific™
25003-104630 |
100 mm | 4.6 mm | 3 μm |
£705.88
Each |
In Stock
Log in to see stock. |
|||
10704556
|
Thermo Scientific™
25003-151030 |
150 mm | 1 mm | 3 μm |
£905.49
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10121223
|
Thermo Scientific™
25003-152130 |
150 mm | 2.1 mm | 3 μm |
£779.33
Each |
In Stock
Log in to see stock. |
|||
10131383
|
Thermo Scientific™
25003-153030 |
150 mm | 3 mm | 3 μm |
£779.33
Each |
In Stock
Log in to see stock. |
|||
10456844
|
Thermo Scientific™
25003-154030 |
150 mm | 4 mm | 3 μm |
£742.12
Each |
In Stock
Log in to see stock. |
|||
10610564
|
Thermo Scientific™
25003-154630 |
150 mm | 4.6 mm | 3 μm |
£742.12
Each |
In Stock
Log in to see stock. |
|||
11528270
|
Thermo Scientific™
25003-014001 |
10 mm | 4 mm | 3 μm |
£392.05
Pack of 4 |
In Stock
Log in to see stock. |
|||
11808901
|
Thermo Scientific™
25002-012135 |
10 mm | 2.1 mm | 1.9 μm |
£627.66
Pack of 3 |
In Stock
Log in to see stock. |
|||
10464613
|
Thermo Scientific™
25002-022130 |
20 mm | 2.1 mm | 1.9 μm |
£751.66
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||
10305063
|
Thermo Scientific™
25002-031030 |
30 mm | 1 mm | 1.9 μm |
£769.59
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10049142
|
Thermo Scientific™
25002-032130 |
30 mm | 2.1 mm | 1.9 μm |
£792.67
Each |
In Stock
Log in to see stock. |
|||
10020213
|
Thermo Scientific™
25002-051030 |
50 mm | 1 mm | 1.9 μm |
£809.41
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
10474783
|
Thermo Scientific™
25002-052130 |
50 mm | 2.1 mm | 1.9 μm |
£805.08
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10345203
|
Thermo Scientific™
25002-053030 |
50 mm | 3 mm | 1.9 μm |
£809.41
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
10169344
|
Thermo Scientific™
25002-054630 |
50 mm | 4.6 mm | 1.9 μm |
£751.66
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
10715004
|
Thermo Scientific™
25002-101030 |
100 mm | 1 mm | 1.9 μm |
£833.69
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10734604
|
Thermo Scientific™
25002-102130 |
100 mm | 2.1 mm | 1.9 μm |
£805.08
Each |
In Stock
Log in to see stock. |
|||
10582465
|
Thermo Scientific™
25002-103030 |
100 mm | 3 mm | 1.9 μm |
£833.69
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
10630204
|
Thermo Scientific™
25002-152130 |
150 mm | 2.1 mm | 1.9 μm |
£915.73
Each |
In Stock
Log in to see stock. |
|||
10446503
|
Thermo Scientific™
25002-202130 |
200 mm | 2.1 mm | 1.9 μm |
£907.14
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10608705
|
Thermo Scientific™
25003-011001 |
10 mm | 1 mm | 3 μm |
£392.05
Pack of 4 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
11518270
|
Thermo Scientific™
25003-012101 |
10 mm | 2.1 mm | 3 μm |
£392.05
Pack of 4 |
In Stock
Log in to see stock. |
|||
10481365
|
Thermo Scientific™
25003-013001 |
10 mm | 3 mm | 3 μm |
£392.05
Pack of 4 |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10618705
|
Thermo Scientific™
25005-011001 |
10 mm | 1 mm | 5 μm |
£380.83
Pack of 4 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
11538270
|
Thermo Scientific™
25005-012101 |
10 mm | 2.1 mm | 5 μm |
£356.76
Pack of 4 |
In Stock
Log in to see stock. |
|||
10544085
|
Thermo Scientific™
25005-013001 |
10 mm | 3 mm | 5 μm |
£356.76
Pack of 4 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
11548270
|
Thermo Scientific™
25005-014001 |
10 mm | 4 mm | 5 μm |
£356.76
Pack of 4 |
In Stock
Log in to see stock. |
|||
10107604
|
Thermo Scientific™
25005-019023 |
10 mm | 10 mm | 5 μm | N/A | ||||
10475163
|
Thermo Scientific™
25005-032130 |
30 mm | 2.1 mm | 5 μm | N/A | ||||
10668825
|
Thermo Scientific™
25005-033030 |
30 mm | 3 mm | 5 μm | N/A | ||||
10008862
|
Thermo Scientific™
25005-034630 |
30 mm | 4.6 mm | 5 μm | N/A | ||||
10611334
|
Thermo Scientific™
25005-052130 |
50 mm | 2.1 mm | 5 μm |
£617.16
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10191523
|
Thermo Scientific™
25005-053030 |
50 mm | 3 mm | 5 μm |
£611.45
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10611524
|
Thermo Scientific™
25005-054630 |
50 mm | 4.6 mm | 5 μm |
£587.59
Each |
In Stock
Log in to see stock. |
|||
10322725
|
Thermo Scientific™
25005-059070 |
50 mm | 10 mm | 5 μm | N/A | ||||
10332485
|
Thermo Scientific™
25005-059270 |
50 mm | 21.2 mm | 5 μm | N/A | ||||
10037444
|
Thermo Scientific™
25005-059370 |
50 mm | 30 mm | 5 μm | N/A | ||||
10517825
|
Thermo Scientific™
25005-059570 |
50 mm | 50 mm | 5 μm | N/A | ||||
10140743
|
Thermo Scientific™
25005-102130 |
100 mm | 2.1 mm | 5 μm |
£667.72
Each |
In Stock
Log in to see stock. |
|||
10384723
|
Thermo Scientific™
|
100 mm | 3 mm | 5 μm |
£702.93
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||
10121033
|
Thermo Scientific™
25005-104630 |
100 mm | 4.6 mm | 5 μm |
£640.05
Each |
In Stock
Log in to see stock. |
|||
10312405
|
Thermo Scientific™
25005-109070 |
100 mm | 10 mm | 5 μm | N/A | ||||
10077404
|
Thermo Scientific™
25005-109270 |
100 mm | 21.2 mm | 5 μm | N/A | ||||
10793216
|
Thermo Scientific™
25005-109370 |
100 mm | 30 mm | 5 μm | N/A | ||||
10087404
|
Thermo Scientific™
25005-109570 |
100 mm | 50 mm | 5 μm | N/A | ||||
10131033
|
Thermo Scientific™
25005-152130 |
150 mm | 2.1 mm | 5 μm |
£689.02
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10795316
|
Thermo Scientific™
25005-153030 |
150 mm | 3 mm | 5 μm |
£689.02
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10313065
|
Thermo Scientific™
25005-154030 |
150 mm | 4 mm | 5 μm |
£676.30
Each |
In Stock
Log in to see stock. |
|||
10501695
|
Thermo Scientific™
25005-154630 |
150 mm | 4.6 mm | 5 μm |
£682.99
Each |
In Stock
Log in to see stock. |
|||
10281195
|
Thermo Scientific™
25005-159070 |
150 mm | 10 mm | 5 μm | N/A | ||||
10437634
|
Thermo Scientific™
25005-159270 |
150 mm | 21.2 mm | 5 μm | N/A | ||||
10027174
|
Thermo Scientific™
25005-159370 |
150 mm | 30 mm | 5 μm | N/A | ||||
10352975
|
Thermo Scientific™
25005-159570 |
150 mm | 50 mm | 5 μm | N/A | ||||
10009102
|
Thermo Scientific™
25005-252130 |
250 mm | 2.1 mm | 5 μm |
£889.96
Each |
In Stock
Log in to see stock. |
|||
10352775
|
Thermo Scientific™
25005-253030 |
250 mm | 3 mm | 5 μm |
£729.77
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10506365
|
Thermo Scientific™
25005-254030 |
250 mm | 4 mm | 5 μm |
£694.43
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10543035
|
Thermo Scientific™
25005-254630 |
250 mm | 4.6 mm | 5 μm |
£695.39
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
10723996
|
Thermo Scientific™
25005-259070 |
250 mm | 10 mm | 5 μm | N/A | ||||
10467634
|
Thermo Scientific™
25005-259270 |
250 mm | 21.2 mm | 5 μm | N/A | ||||
10724186
|
Thermo Scientific™
25005-259370 |
250 mm | 30 mm | 5 μm | N/A | ||||
10372545
|
Thermo Scientific™
25005-259570 |
250 mm | 50 mm | 5 μm | N/A | ||||
10076344
|
Thermo Scientific™
25012-019023 |
10 mm | 10 mm | 12 μm | N/A | ||||
15240718
|
Thermo Scientific™
25012-019223 |
10 mm | 10 mm | 12 μm | N/A | ||||
10578035
|
Thermo Scientific™
25012-059070 |
50 mm | 10 mm | 12 μm | N/A | ||||
10078424
|
Thermo Scientific™
25012-059270 |
50 mm | 21.2 mm | 12 μm | N/A | ||||
10743426
|
Thermo Scientific™
25012-059370 |
50 mm | 30 mm | 12 μm | N/A | ||||
10699775
|
Thermo Scientific™
25012-059570 |
50 mm | 50 mm | 12 μm | N/A | ||||
10392785
|
Thermo Scientific™
25012-109070 |
100 mm | 10 mm | 12 μm | N/A | ||||
10272055
|
Thermo Scientific™
25012-109270 |
100 mm | 21.2 mm | 12 μm | N/A | ||||
10698825
|
Thermo Scientific™
25012-109370 |
100 mm | 30 mm | 12 μm | N/A | ||||
10382405
|
Thermo Scientific™
25012-109570 |
100 mm | 50 mm | 12 μm | N/A | ||||
10417644
|
Thermo Scientific™
25012-159070 |
150 mm | 10 mm | 12 μm | N/A | ||||
10508615
|
Thermo Scientific™
25012-159270 |
150 mm | 21.2 mm | 12 μm | N/A | ||||
10008524
|
Thermo Scientific™
25012-159370 |
150 mm | 30 mm | 12 μm | N/A | ||||
10639405
|
Thermo Scientific™
25012-159570 |
150 mm | 50 mm | 12 μm | N/A | ||||
10363465
|
Thermo Scientific™
25012-259070 |
250 mm | 10 mm | 12 μm | N/A | ||||
10609595
|
Thermo Scientific™
25012-259270 |
250 mm | 21.2 mm | 12 μm | N/A | ||||
11508690
|
Thermo Scientific™
25012-259370 |
250 mm | 30 mm | 12 μm | N/A | ||||
10713046
|
Thermo Scientific™
25012-259570 |
250 mm | 50 mm | 12 μm | N/A | ||||
15260718
|
Thermo Scientific™
854-00 |
10 mm | 20 mm | 12 μm | N/A | ||||
11528360
|
Thermo Scientific™
25003-253030 |
250 mm | 3 mm | 3 μm | N/A | ||||
11827041
|
Thermo Scientific™
25003-254630 |
250 mm | 4.6 mm | 3 μm |
£785.04
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||
Description
Achieve exceptional peak shape and resolution for your HPLC and LC/MS applications with Thermo Scientific™ Hypersil GOLD™ HPLC columns. These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in the accuracy and quality of your analytical data.
Novel Bonding
- Proprietary derivatization and endcapping
- C18 (USP L1) selectivity
pH Stability
- Robust performance at high pH
- Excellent column stability at low pH
Flexible Formats
- Javelin HTS columns for high-throughput screening
- Semi-preparative columns with 12 μm particles available
About the Hypersil GOLD Family of HPLC Columns
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different column chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
- Excellent peak symmetry
- Narrow peaks for outstanding efficiency
- Enhanced sensitivity and improved resolution
- Exceptional reproducibility for reliable chromatography, column after column
- Solutions for high-throughput screening
- 1.9, 3, and 5 μm particle sizes for all your separation needs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.