Learn More
Thermo Scientific™ Hypersil™ MOS-2 C8 HPLC Columns
Use as a general-purpose C8 phase for acidic and neutral analytes, with decrease silanol activity
£330.77 - £595.14
Specifications
Max. Pressure | 5800 psi (400 bar) |
---|---|
pH | 2 to 8 |
Pore Size | 120 Å |
Surface Area | 170 m²/g |
Temperature | 60°C |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10776334
|
Thermo Scientific™
30305-154630 |
5 μm | 4.6 mm | 150 mm |
£571.05
Each |
In Stock
Log in to see stock. |
|||
10744804
|
Thermo Scientific™
30305-254630 |
5 μm | 4.6 mm | 250 mm |
£595.14
Each |
In Stock
Log in to see stock. |
|||
10562075
|
Thermo Scientific™
30305-014001 |
5 μm | 4 mm | 10 mm |
£330.77
Pack of 4 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
10733026
|
Thermo Scientific™
30305-254030 |
5 μm | 4 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS-2 C8 HPLC Columns.
Hypersil stationary phases are recognized as an HPLC industry standard. A proven effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.
For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Manufactured under ISO 9001-2000 quality guidelines
- Affordable
Specifications
5800 psi (400 bar) | |
120 Å | |
60°C | |
C8 | |
6.5% | |
Reversed Phase |
2 to 8 | |
170 m²/g | |
L7 | |
Hypersil Classical | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.