Learn More
Thermo Scientific™ Hypersil™ PREP BDS C18 HPLC Columns
Scale up with Hypersil PREP C18 BDS preparative LC columns for routine applications and C18 retention with base deactivation for reduced silanol activity.
£162.14 - £14051.15
Specifications
pH | 2 to 8 |
---|---|
Pore Size | 145 Å |
Surface Area | 185 m²/g |
Temperature | 60°C |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||
16328856
|
Thermo Scientific™
40003-052130 |
2 mm | 50 mm | 3 μm | Preparative Column, Standard |
£505.05
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
16338856
|
Thermo Scientific™
40003-054630 |
4.6 mm | 50 mm | 3 μm | Preparative Column, Standard |
£446.40
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16348856
|
Thermo Scientific™
40003-102130 |
2 mm | 100 mm | 3 μm | Preparative Column, Standard |
£550.78
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16358856
|
Thermo Scientific™
40003-104630 |
4.6 mm | 100 mm | 3 μm | Preparative Column, Standard |
£547.80
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
16368856
|
Thermo Scientific™
40003-152130 |
2 mm | 150 mm | 3 μm | Preparative Column, Standard |
£613.42
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16378856
|
Thermo Scientific™
40003-154630 |
4.6 mm | 150 mm | 3 μm | Preparative Column, Standard |
£615.41
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
16388856
|
Thermo Scientific™
40003-252130 |
2 mm | 250 mm | 3 μm | Preparative Column, Standard |
£652.19
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16398856
|
Thermo Scientific™
40003-254630 |
4.6 mm | 250 mm | 3 μm | Preparative Column, Standard |
£624.35
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||
16308866
|
Thermo Scientific™
40005-052130 |
2 mm | 50 mm | 5 μm | Preparative Column, Standard |
£434.47
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16318866
|
Thermo Scientific™
40005-054630 |
4.6 mm | 50 mm | 5 μm | Preparative Column, Standard |
£457.33
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16328866
|
Thermo Scientific™
40005-059070A |
10 mm | 50 mm | 5 μm | Preparative Column, Standard |
£930.56
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16338866
|
Thermo Scientific™
40005-059270A |
20 mm | 50 mm | 5 μm | Preparative Column, Standard |
£1085.66
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16348866
|
Thermo Scientific™
40005-059270XL |
25 mm | 50 mm | 5 μm | Preparative Column, XtendedLife |
£3829.50
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16358866
|
Thermo Scientific™
40005-102130 |
2 mm | 100 mm | 5 μm | Preparative Column, Standard |
£561.72
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16368866
|
Thermo Scientific™
40005-104630 |
4.6 mm | 100 mm | 5 μm | Preparative Column, Standard |
£492.13
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16378866
|
Thermo Scientific™
40005-109070A |
10 mm | 100 mm | 5 μm | Preparative Column, Standard |
£962.38
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16388866
|
Thermo Scientific™
40005-109270A |
20 mm | 100 mm | 5 μm | Preparative Column, Standard |
£2399.99
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16398866
|
Thermo Scientific™
40005-109270XL |
25 mm | 100 mm | 5 μm | Preparative Column, XtendedLife |
£5188.48
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
16308876
|
Thermo Scientific™
40005-152130 |
2 mm | 150 mm | 5 μm | Preparative Column, Standard |
£530.90
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16318876
|
Thermo Scientific™
40005-154630 |
4.6 mm | 150 mm | 5 μm | Preparative Column, Standard |
£547.80
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16328876
|
Thermo Scientific™
40005-159070A |
10 mm | 150 mm | 5 μm | Preparative Column, Standard |
£1118.47
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
16338876
|
Thermo Scientific™
40005-159270A |
20 mm | 150 mm | 5 μm | Preparative Column, Standard |
£2730.05
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16348876
|
Thermo Scientific™
40005-159270XL |
25 mm | 150 mm | 5 μm | Preparative Column, XtendedLife |
£5011.42
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16358876
|
Thermo Scientific™
40005-159370A |
30 mm | 150 mm | 5 μm | Preparative Column, Standard |
£4455.97
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16368876
|
Thermo Scientific™
40005-159370XL |
30 mm | 150 mm | 5 μm | Preparative Column, XtendedLife |
£7344.18
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16378876
|
Thermo Scientific™
40005-159570A |
50 mm | 150 mm | 5 μm | Preparative Column, Standard |
£9786.83
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16388876
|
Thermo Scientific™
40005-159570XL |
50 mm | 150 mm | 5 μm | Preparative Column, XtendedLife |
£13037.27
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16398876
|
Thermo Scientific™
40005-252130 |
2 mm | 250 mm | 5 μm | Preparative Column, Standard |
£567.69
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16308886
|
Thermo Scientific™
40005-254630 |
4.6 mm | 250 mm | 5 μm | Preparative Column, Standard |
£554.76
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||
16318886
|
Thermo Scientific™
40005-259070A |
10 mm | 250 mm | 5 μm | Preparative Column, Standard |
£1268.30
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
16328886
|
Thermo Scientific™
40005-259270A |
20 mm | 250 mm | 5 μm | Preparative Column, Standard |
£3070.06
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||
16338886
|
Thermo Scientific™
40005-259270XL |
25 mm | 250 mm | 5 μm | Preparative Column, XtendedLife |
£5711.13
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16348886
|
Thermo Scientific™
40005-259370A |
30 mm | 250 mm | 5 μm | Preparative Column, Standard |
£5324.37
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
16358886
|
Thermo Scientific™
40005-259370XL |
30 mm | 250 mm | 5 μm | Preparative Column, XtendedLife |
£8054.51
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16368886
|
Thermo Scientific™
40005-259570A |
50 mm | 250 mm | 5 μm | Preparative Column, Standard |
£10792.96
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16378886
|
Thermo Scientific™
40005-259570XL |
50 mm | 250 mm | 5 μm | Preparative Column, XtendedLife |
£14051.15
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16388886
|
Thermo Scientific™
40010-018823A |
10 mm | 8 mm | 10 μm | Guard Column |
£162.14
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
16398886
|
Thermo Scientific™
40010-019023A |
10 mm | 20 mm | 10 μm | Guard Column |
£211.39
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||
16308896
|
Thermo Scientific™
40010-019323A |
10 mm | 30 mm | 10 μm | Guard 1/16 in. |
£256.16
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16318896
|
Thermo Scientific™
40010-019323B |
10 mm | 30 mm | 10 μm | Guard 1/8 in. |
£256.16
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16338896
|
Thermo Scientific™
40010-029323A |
20 mm | 30 mm | 10 μm | Guard 1/16 in. |
£422.77
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16348896
|
Thermo Scientific™
40010-029323B |
20 mm | 30 mm | 10 μm | Guard 1/8 in. |
£422.77
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16358896
|
Thermo Scientific™
40010-059070A |
10 mm | 50 mm | 10 μm | Preparative Column, Standard |
£837.12
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16368896
|
Thermo Scientific™
40010-059270A |
20 mm | 50 mm | 10 μm | Preparative Column, Standard |
£976.30
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16378896
|
Thermo Scientific™
40010-059270XL |
25 mm | 50 mm | 10 μm | Preparative Column, XtendedLife |
£3194.42
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16388896
|
Thermo Scientific™
40010-109070A |
10 mm | 100 mm | 10 μm | Preparative Column, Standard |
£865.94
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||
16398896
|
Thermo Scientific™
40010-109270A |
20 mm | 100 mm | 10 μm | Preparative Column, Standard |
£2159.38
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16308906
|
Thermo Scientific™
40010-109270XL |
25 mm | 100 mm | 10 μm | Preparative Column, XtendedLife |
£4502.37
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16318906
|
Thermo Scientific™
40010-159070A |
10 mm | 150 mm | 10 μm | Preparative Column, Standard |
£1005.14
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
16328906
|
Thermo Scientific™
40010-159270A |
20 mm | 150 mm | 10 μm | Preparative Column, Standard |
£2457.65
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16338906
|
Thermo Scientific™
40010-159270XL |
25 mm | 150 mm | 10 μm | Preparative Column, XtendedLife |
£4816.92
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16348906
|
Thermo Scientific™
40010-159370A |
30 mm | 150 mm | 10 μm | Preparative Column, Standard |
£4009.59
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16358906
|
Thermo Scientific™
40010-159370XL |
30 mm | 150 mm | 10 μm | Preparative Column, XtendedLife |
£6882.98
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16368906
|
Thermo Scientific™
40010-159570A |
50 mm | 150 mm | 10 μm | Preparative Column, Standard |
£8809.54
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16378906
|
Thermo Scientific™
40010-159570XL |
50 mm | 150 mm | 10 μm | Preparative Column, XtendedLife |
£12054.42
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16388906
|
Thermo Scientific™
40010-259070A |
10 mm | 250 mm | 10 μm | Preparative Column, Standard |
£1140.64
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16398906
|
Thermo Scientific™
40010-259270A |
20 mm | 250 mm | 10 μm | Preparative Column, Standard |
£2764.85
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16308916
|
Thermo Scientific™
40010-259270XL |
25 mm | 250 mm | 10 μm | Preparative Column, XtendedLife |
£4874.53
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
16318916
|
Thermo Scientific™
40010-259370A |
30 mm | 250 mm | 10 μm | Preparative Column, Standard |
£4793.16
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16328916
|
Thermo Scientific™
40010-259370XL |
30 mm | 250 mm | 10 μm | Preparative Column, XtendedLife |
£7314.32
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16338916
|
Thermo Scientific™
40010-259570A |
50 mm | 250 mm | 10 μm | Preparative Column, Standard |
£9714.26
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16348916
|
Thermo Scientific™
40010-259570XL |
50 mm | 250 mm | 10 μm | Preparative Column, XtendedLife |
£12964.21
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16328896
|
Thermo Scientific™
40010-029023A |
20 mm | 20 mm | 10 μm | Guard Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Easily scale up your analytical method using Thermo Scientific™ Hypersil™ PREP BDS C18 reversed phase preparative HPLC Columns. Our prep columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. These endcapped, silica-based columns offer C18 retention with base deactivation for reduced silanol activity and are an excellent choices for routine applications which require development from analytical to preparative LC scale.
Excellent Choice for Routine Applications
- C18 (USP L1) selectivity.
- Base deactivated silica.
- pH stable from 2 to 8.
- Maximum temperature of 60°C.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
- Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
- Standard threaded hardware for column i.d. < 40mm.
- Standard flange hardware for column i.d. ≥ 40mm.
- Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
- Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
- Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
- If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
- Please contact your local sales representative for Customer Specific Orders.
- Custom built columns are not eligible for returns.
Specifications
2 to 8 | |
185 m²/g | |
L1 | |
Spherical, Fully Porous, Base-Deactivated Silica | |
Reversed Phase | |
Reversed Phase |
145 Å | |
60°C | |
C18 | |
11% | |
Yes |