Learn More
Thermo Scientific™ Hypersil™ PREP HS C18 HPLC Columns
Maximize retention and loading with Hypersil PREP HS C18 prep LC reversed phase columns which offers high loading capacity and suitable for USP-L1 Applications.
£164.46 - £13685.65
Specifications
pH | 2 to 9 |
---|---|
Pore Size | 100 Å |
Surface Area | 350 m²/g |
Temperature | 60°C |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||
16498403
|
Thermo Scientific™
41003-052130 |
2 mm | 50 mm | 3 μm | Preparative Column, Standard |
£401.65
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16408413
|
Thermo Scientific™
41003-054630 |
4.6 mm | 50 mm | 3 μm | Preparative Column, Standard |
£446.40
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16418413
|
Thermo Scientific™
41003-102130 |
2 mm | 100 mm | 3 μm | Preparative Column, Standard |
£503.06
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||
16428413
|
Thermo Scientific™
41003-104630 |
4.6 mm | 100 mm | 3 μm | Preparative Column, Standard |
£547.80
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16438413
|
Thermo Scientific™
41003-152130 |
2 mm | 150 mm | 3 μm | Preparative Column, Standard |
£558.74
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16448413
|
Thermo Scientific™
41003-154630 |
4.6 mm | 150 mm | 3 μm | Preparative Column, Standard |
£615.41
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||
16458413
|
Thermo Scientific™
41003-252130 |
2 mm | 250 mm | 3 μm | Preparative Column, Standard |
£652.19
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16468413
|
Thermo Scientific™
41003-254630 |
4.6 mm | 250 mm | 3 μm | Preparative Column, Standard |
£676.05
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||
16478413
|
Thermo Scientific™
41005-052130 |
2 mm | 50 mm | 5 μm | Preparative Column, Standard |
£344.99
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16488413
|
Thermo Scientific™
41005-054630 |
4.6 mm | 50 mm | 5 μm | Preparative Column, Standard |
£399.67
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16498413
|
Thermo Scientific™
41005-059070A |
10 mm | 50 mm | 5 μm | Preparative Column, Standard |
£941.50
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16408423
|
Thermo Scientific™
41005-059270A |
20 mm | 50 mm | 5 μm | Preparative Column, Standard |
£997.18
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16418423
|
Thermo Scientific™
41005-059270XL |
25 mm | 50 mm | 5 μm | Preparative Column, XtendedLife |
£3734.85
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16428423
|
Thermo Scientific™
41005-102130 |
2 mm | 100 mm | 5 μm | Preparative Column, Standard |
£446.40
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16438423
|
Thermo Scientific™
41005-104630 |
4.6 mm | 100 mm | 5 μm | Preparative Column, Standard |
£492.13
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16448423
|
Thermo Scientific™
41005-109070A |
10 mm | 100 mm | 5 μm | Preparative Column, Standard |
£997.18
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16458423
|
Thermo Scientific™
41005-109270A |
20 mm | 100 mm | 5 μm | Preparative Column, Standard |
£1882.01
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||
16468423
|
Thermo Scientific™
41005-109270XL |
25 mm | 100 mm | 5 μm | Preparative Column, XtendedLife |
£4965.43
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16478423
|
Thermo Scientific™
41005-152130 |
2 mm | 150 mm | 5 μm | Preparative Column, Standard |
£484.17
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16488423
|
Thermo Scientific™
41005-154630 |
4.6 mm | 150 mm | 5 μm | Preparative Column, Standard |
£547.80
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16498423
|
Thermo Scientific™
41005-159070A |
10 mm | 150 mm | 5 μm | Preparative Column, Standard |
£1140.34
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16408433
|
Thermo Scientific™
41005-159270A |
20 mm | 150 mm | 5 μm | Preparative Column, Standard |
£2250.85
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16418433
|
Thermo Scientific™
41005-159270XL |
25 mm | 150 mm | 5 μm | Preparative Column, XtendedLife |
£5426.11
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16428433
|
Thermo Scientific™
41005-159370A |
30 mm | 150 mm | 5 μm | Preparative Column, Standard |
£3679.50
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16438433
|
Thermo Scientific™
41005-159370XL |
30 mm | 150 mm | 5 μm | Preparative Column, XtendedLife |
£6720.32
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16448433
|
Thermo Scientific™
41005-159570A |
50 mm | 150 mm | 5 μm | Preparative Column, Standard |
£7337.14
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16458433
|
Thermo Scientific™
41005-159570XL |
50 mm | 150 mm | 5 μm | Preparative Column, XtendedLife |
£11579.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16468433
|
Thermo Scientific™
41005-252130 |
2 mm | 250 mm | 5 μm | Preparative Column, Standard |
£567.69
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16478433
|
Thermo Scientific™
41005-254630 |
4.6 mm | 250 mm | 5 μm | Preparative Column, Standard |
£652.19
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
16488433
|
Thermo Scientific™
41005-259070A |
10 mm | 250 mm | 5 μm | Preparative Column, Standard |
£1253.68
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16498433
|
Thermo Scientific™
41005-259270A |
20 mm | 250 mm | 5 μm | Preparative Column, Standard |
£2708.18
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
16408443
|
Thermo Scientific™
41005-259270XL |
25 mm | 250 mm | 5 μm | Preparative Column, XtendedLife |
£5312.12
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16418443
|
Thermo Scientific™
41005-259370A |
30 mm | 250 mm | 5 μm | Preparative Column, Standard |
£4369.03
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16428443
|
Thermo Scientific™
41005-259370XL |
30 mm | 250 mm | 5 μm | Preparative Column, XtendedLife |
£7099.16
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16438443
|
Thermo Scientific™
41005-259570A |
50 mm | 250 mm | 5 μm | Preparative Column, Standard |
£8254.79
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
16448443
|
Thermo Scientific™
41005-259570XL |
50 mm | 250 mm | 5 μm | Preparative Column, XtendedLife |
£13685.65
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16458443
|
Thermo Scientific™
41010-018823A |
10 mm | 8 mm | 10 μm | Guard Column |
£164.46
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16468443
|
Thermo Scientific™
41010-019023A |
10 mm | 20 mm | 10 μm | Guard Column |
£211.39
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
16478443
|
Thermo Scientific™
41010-019323A |
10 mm | 30 mm | 10 μm | Guard 1/16 in. |
£256.16
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16488443
|
Thermo Scientific™
41010-019323B |
10 mm | 30 mm | 10 μm | Guard 1/8 in. |
£256.16
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16408453
|
Thermo Scientific™
41010-029323A |
20 mm | 30 mm | 10 μm | Guard 1/16 in. |
£422.77
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16418453
|
Thermo Scientific™
41010-029323B |
20 mm | 30 mm | 10 μm | Guard 1/8 in. |
£422.77
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16428453
|
Thermo Scientific™
41010-059070A |
10 mm | 50 mm | 10 μm | Preparative Column, Standard |
£884.83
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16438453
|
Thermo Scientific™
41010-059270A |
20 mm | 50 mm | 10 μm | Preparative Column, Standard |
£911.68
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16448453
|
Thermo Scientific™
41010-059270XL |
25 mm | 50 mm | 10 μm | Preparative Column, XtendedLife |
£3854.19
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16458453
|
Thermo Scientific™
41010-109070A |
10 mm | 100 mm | 10 μm | Preparative Column, Standard |
£941.50
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16468453
|
Thermo Scientific™
41010-109270A |
20 mm | 100 mm | 10 μm | Preparative Column, Standard |
£1131.39
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16478453
|
Thermo Scientific™
41010-109270XL |
25 mm | 100 mm | 10 μm | Preparative Column, XtendedLife |
£4369.39
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16488453
|
Thermo Scientific™
41010-159070A |
10 mm | 150 mm | 10 μm | Preparative Column, Standard |
£1083.67
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
16498453
|
Thermo Scientific™
41010-159270A |
20 mm | 150 mm | 10 μm | Preparative Column, Standard |
£2194.18
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16408463
|
Thermo Scientific™
41010-159270XL |
25 mm | 150 mm | 10 μm | Preparative Column, XtendedLife |
£4955.86
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16418463
|
Thermo Scientific™
41010-159370A |
30 mm | 150 mm | 10 μm | Preparative Column, Standard |
£4009.59
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16428463
|
Thermo Scientific™
41010-159370XL |
30 mm | 150 mm | 10 μm | Preparative Column, XtendedLife |
£7329.10
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16438463
|
Thermo Scientific™
41010-159570A |
50 mm | 150 mm | 10 μm | Preparative Column, Standard |
£7337.14
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16448463
|
Thermo Scientific™
41010-159570XL |
50 mm | 150 mm | 10 μm | Preparative Column, XtendedLife |
£10572.14
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16458463
|
Thermo Scientific™
41010-259070A |
10 mm | 250 mm | 10 μm | Preparative Column, Standard |
£1239.47
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16468463
|
Thermo Scientific™
41010-259270A |
20 mm | 250 mm | 10 μm | Preparative Column, Standard |
£2622.68
Each |
In Stock
Log in to see stock. |
||
16478463
|
Thermo Scientific™
41010-259270XL |
25 mm | 250 mm | 10 μm | Preparative Column, XtendedLife |
£4900.24
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16488463
|
Thermo Scientific™
41010-259370A |
30 mm | 250 mm | 10 μm | Preparative Column, Standard |
£4064.31
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
16498463
|
Thermo Scientific™
41010-259370XL |
30 mm | 250 mm | 10 μm | Preparative Column, XtendedLife |
£6653.13
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16408473
|
Thermo Scientific™
41010-259570A |
50 mm | 250 mm | 10 μm | Preparative Column, Standard |
£8025.13
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16418473
|
Thermo Scientific™
41010-259570XL |
50 mm | 250 mm | 10 μm | Preparative Column, XtendedLife |
£11264.74
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16498443
|
Thermo Scientific™
41010-029023A |
20 mm | 20 mm | 10 μm | Guard Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Maximize retention and loading using Thermo Scientific™ Hypersil™ PREP HS C18 reversed phase preparative HPLC Columns. Our columns come in a variety of dimensions allowing you to easily scale up your analytical method, including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. These silica-based columns are packed with high surface area particles with a 100Å pore size offering C18 retention, an excellent choice for small molecule and drug purification applications.
Excellent Choice for Routine Applications
- C18 (USP L1) selectivity - high surface area particle with 100Å pore size.
- pH stable from 2 to 9.
- Maximum temperature of 60°C.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
- Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
- Standard threaded hardware for column i.d. < 40mm.
- Standard flange hardware for column i.d. ≥ 40mm.
- Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
- Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
- Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
- If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
- Please contact your local sales representative for Customer Specific Orders.
- Custom built columns are not eligible for returns.
Specifications
2 to 9 | |
350 m²/g | |
L1 | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
100 Å | |
60°C | |
C18 | |
15% | |
Yes |