Learn More
Thermo Scientific™ Hypersil™ PREP Si HPLC Columns
Separate nonpolar and moderately polar organic compounds with Hypersil PREP Si prep LC columns for normal phase methods, suitable for USP L3 applications.
£120.80 - £13917.33
Specifications
Particle Size | 5 μm |
---|---|
pH | 2 to 7 |
Pore Size | 130 Å |
Surface Area | 195 m²/g |
Temperature | 60°C |
Product Code | Brand | Column Format | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Format | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
16349106
|
Thermo Scientific™
40405-018823A |
Guard Column | 10 mm | 8 mm |
£120.80
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16359106
|
Thermo Scientific™
40405-019023A |
Guard Column | 10 mm | 20 mm |
£211.39
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16369106
|
Thermo Scientific™
40405-019323A |
Guard 1/16 in. | 10 mm | 30 mm |
£256.16
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16379106
|
Thermo Scientific™
40405-019323B |
Guard 1/8 in. | 10 mm | 30 mm |
£256.16
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16399106
|
Thermo Scientific™
40405-029323A |
Guard 1/16 in. | 20 mm | 30 mm |
£422.77
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16309116
|
Thermo Scientific™
40405-029323B |
Guard 1/8 in. | 20 mm | 30 mm |
£422.77
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16319116
|
Thermo Scientific™
40405-052130 |
Preparative Column, Standard | 2 mm | 50 mm |
£434.47
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16329116
|
Thermo Scientific™
40405-054630 |
Preparative Column, Standard | 4.6 mm | 50 mm |
£457.33
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16339116
|
Thermo Scientific™
40405-059070A |
Preparative Column, Standard | 10 mm | 50 mm |
£930.56
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16349116
|
Thermo Scientific™
40405-059270A |
Preparative Column, Standard | 20 mm | 50 mm |
£1085.66
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16359116
|
Thermo Scientific™
40405-059270XL |
Preparative Column, XtendedLife | 25 mm | 50 mm |
£3829.50
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16369116
|
Thermo Scientific™
40405-102130 |
Preparative Column, Standard | 2 mm | 100 mm |
£561.72
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16379116
|
Thermo Scientific™
40405-104630 |
Preparative Column, Standard | 4.6 mm | 100 mm |
£492.13
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16389116
|
Thermo Scientific™
40405-109070A |
Preparative Column, Standard | 10 mm | 100 mm |
£962.38
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16399116
|
Thermo Scientific™
40405-109270A |
Preparative Column, Standard | 20 mm | 100 mm |
£2399.99
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16309126
|
Thermo Scientific™
40405-109270XL |
Preparative Column, XtendedLife | 25 mm | 100 mm |
£5188.48
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16319126
|
Thermo Scientific™
40405-152130 |
Preparative Column, Standard | 2 mm | 150 mm |
£530.90
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16329126
|
Thermo Scientific™
40405-154630 |
Preparative Column, Standard | 4.6 mm | 150 mm |
£547.80
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16339126
|
Thermo Scientific™
40405-159070A |
Preparative Column, Standard | 10 mm | 150 mm |
£1118.47
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16349126
|
Thermo Scientific™
40405-159270A |
Preparative Column, Standard | 20 mm | 150 mm |
£2730.05
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16359126
|
Thermo Scientific™
40405-159270XL |
Preparative Column, XtendedLife | 25 mm | 150 mm |
£5800.25
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16369126
|
Thermo Scientific™
40405-159370A |
Preparative Column, Standard | 30 mm | 150 mm |
£4455.97
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16379126
|
Thermo Scientific™
40405-159370XL |
Preparative Column, XtendedLife | 30 mm | 150 mm |
£7344.18
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16389126
|
Thermo Scientific™
40405-159570A |
Preparative Column, Standard | 50 mm | 150 mm |
£9786.83
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16399126
|
Thermo Scientific™
40405-159570XL |
Preparative Column, XtendedLife | 50 mm | 150 mm |
£12913.10
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16309136
|
Thermo Scientific™
40405-252130 |
Preparative Column, Standard | 2 mm | 250 mm |
£567.69
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16319136
|
Thermo Scientific™
40405-254630 |
Preparative Column, Standard | 4.6 mm | 250 mm |
£554.76
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16329136
|
Thermo Scientific™
40405-259070A |
Preparative Column, Standard | 10 mm | 250 mm |
£1268.30
Each |
In Stock
Log in to see stock. |
|||
16339136
|
Thermo Scientific™
40405-259270A |
Preparative Column, Standard | 20 mm | 250 mm |
£3070.06
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16349136
|
Thermo Scientific™
40405-259270XL |
Preparative Column, XtendedLife | 25 mm | 250 mm |
£5711.13
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16359136
|
Thermo Scientific™
40405-259370A |
Preparative Column, Standard | 30 mm | 250 mm |
£5324.37
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16369136
|
Thermo Scientific™
40405-259370XL |
Preparative Column, XtendedLife | 30 mm | 250 mm |
£8054.51
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16379136
|
Thermo Scientific™
40405-259570A |
Preparative Column, Standard | 50 mm | 250 mm |
£10792.96
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16389136
|
Thermo Scientific™
40405-259570XL |
Preparative Column, XtendedLife | 50 mm | 250 mm |
£13917.33
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
16389106
|
Thermo Scientific™
40405-029023A |
Guard Column | 20 mm | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Use Thermo Scientific™ Hypersil™ PREP Si preparative HPLC columns to scale up your normal phase chromatography analytical method. The silica chemistry separates nonpolar and moderately polar organic compounds. Our preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Normal phase chromatography
- Silica to separate nonpolar and moderately polar organic compounds.
- USP L3.
- pH stable from 2 to 7.
- Maximum temperature of 60°C.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
- Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
- Standard threaded hardware for column i.d. < 40mm.
- Standard flange hardware for column i.d. ≥ 40mm.
- Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
- Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
- Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
- If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
- Please contact your local sales representative for Customer Specific Orders.
- Custom built columns are not eligible for returns.
Specifications
5 μm | |
130 Å | |
60°C | |
Silica | |
0% | |
No |
2 to 7 | |
195 m²/g | |
L3 | |
Spherical, Fully Porous Silica | |
Normal Phase | |
Normal Phase |