missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Hypo-Vial™ Sample Storage Vials
Thermo Scientific™ Hypo-Vial Sample Storage Vials are available in two sizes for storage, heating and autoclaving.
Brand: Thermo Scientific™ TS-12995
This item is not returnable.
View return policy
Description
Thermo Scientific™ Hypo-Vial Sample Storage Vials are available in two sizes for storage, heating and autoclaving.
- Made of high quality borosilicate glass for heating or autoclaving 20mm thick
- Instant syringe access to completely sealed reagents and standards
Specifications
Clear Sample Storage Vial; 125mL | |
Glass | |
125 mL |