missing translation for 'onlineSavingsMsg'
Learn More

Hypophosphorous Acid Solution, Honeywell

Product Code. 15631970
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6303-21-5
Molecular Formula H3O2P
Molecular Weight (g/mol) 66.00
MDL Number MFCD02183592
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
Synonym hypophosphite ion, hypophosphorus acid, phosphinic acid, ion 1-, hydroxyphosphanone, oxo-, e5-phosphanol, hydroxidooxidophosphorus, perfluoroalkyl c6-c12 phosphonic acid, diperfluoroalkyl c6-c12 phosphinic acid, hydroxy-, e5-phosphanone, perfluoro-c6-c12 ethyl hydrogen phosphate
PubChem CID 22497
ChEBI CHEBI:36363
IUPAC Name phosphinic acid
SMILES O[PH2]=O

Specifications

Concentration 50wt. % in H2O
Density 1.206g/mL at 25°C
Vapor Pressure 17mmHg (20°C)
Quantity 2.5 L
Linear Formula H3PO2
UN Number 3264
Formula Weight 66g/mol
Chemical Name or Material Hypophosphorous Acid Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'