Learn More
Hypoxanthine, 99%, Thermo Scientific Chemicals
£20.70 - £141.00
Chemical Identifiers
CAS | 68-94-0 |
---|---|
Molecular Formula | C5H4N4O |
Molecular Weight (g/mol) | 136.11 |
MDL Number | MFCD00005725 |
InChI Key | FDGQSTZJBFJUBT-UHFFFAOYSA-N |
Synonym | hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one |
PubChem CID | 790 |
ChEBI | CHEBI:17368 |
IUPAC Name | 3,7-dihydropurin-6-one |
SMILES | O=C1N=CNC2=C1NC=N2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460373
|
Thermo Scientific Alfa Aesar
A11481.06 |
5 g |
£20.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470373
|
Thermo Scientific Alfa Aesar
A11481.14 |
25 g |
£41.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480373
|
Thermo Scientific Alfa Aesar
A11481.22 |
100 g |
£141.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol A (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol å (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.
Solubility
It does not mix appreciably with water (1:1400). Small octahedra crystals from water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions. Sterilize by filtration through a 0.22 µm filter.
Chemical Identifiers
68-94-0 | |
136.11 | |
FDGQSTZJBFJUBT-UHFFFAOYSA-N | |
790 | |
3,7-dihydropurin-6-one |
C5H4N4O | |
MFCD00005725 | |
hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one | |
CHEBI:17368 | |
O=C1N=CNC2=C1NC=N2 |
Specifications
68-94-0 | |
C5H4N4O | |
5811 | |
hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one | |
FDGQSTZJBFJUBT-UHFFFAOYSA-N | |
3,7-dihydropurin-6-one | |
790 | |
136.11 | |
Hypoxanthine |
>300°C | |
MFCD00005725 | |
14,4869 | |
It does not mix appreciably with water (1:1400). Small octahedra crystals from water. | |
O=C1N=CNC2=C1NC=N2 | |
136.11 | |
CHEBI:17368 | |
99% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 200-697-3
missing translation for 'rtecsNumber' : UP0791000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only