missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyQ™-RS Reduced Serum Media

Product Code. p-7058052
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
This item is not returnable. View return policy

Product Code. 10123492

Brand: Cytiva SH30564.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For applications requiring a lower concentration of serum

  • Support cell culture growth
  • Format: Liquid
  • Store at 2° to 8°C
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Reduced Serum Medium
Sterilization Method 0.1 μm Sterile-filtered
With Additives Glutamine, Glucose
Quantity 500 mL
Content And Storage 2° to 8°C
Description MEM-RS with L-glutamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.