missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyrTrp Cell Dissociation Reagent

Recombinant and animal-derived component-free alternative to traditional porcine trypsin

Brand:  Cytiva SV30209.02

78.82 GBP valid until 2024-12-31
Use promo code "21945" to get your promotional price.


 View more versions of this product

Product Code. 17215814

This item is not returnable. View return policy

Description

Description

  • Animal-derived component-free (ADCF)
  • Dye-free
  • Suitable for a variety of applications and cell types
  • Effective cell dissociation
  • Does not impact cell line viability and doubling times
  • For cell-based vaccine production and stem cell/primary cell/gene therapy cell culture processes
  • For use with attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum
TRUSTED_SUSTAINABILITY
Specifications

Specifications

-20 °C
Cell Culture Dissociation Reagent
Attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers