Learn More
BUCHI I-180 Controller for Rotavapor R-80
The I-180 is an easy-to-use interface for conveniently regulating your vacuum automatically. Increase control of the evaporation process by setting the desired vacuum and accessing other modes of operation.
Brand: BUCHI 11I1803
This item is not returnable.
View return policy
Description
- Functional design fulfills essential needs of vacuum regulation
- Saves resources thanks to control of vacuum pump
- High longevity due to robust and durable design
- User-friendly and intuitive handling due to self-explaining user interface
- Easy and accurate vacuum regulation
- Relevant information is displayed at a glance
- High reliability due to centralized control of major process parameters
- Full process control thanks to parameter monitoring
- Proven technology for major applications
Specifications
Controller Interface | |
Rotavapor R-80 I-100 | |
Adapter Cable for I-180, I-100 Power Supply |
Product Content Correction
Product Title
BUCHI I-180 Controller for Rotavapor R-80 > I-180 Controller for Rotavapor R-80; For Use With (Equipment): Rotavapor R-80 I-100; Includes: Adapter Cable for I-180, I-100 Power Supply
Spot an opportunity for improvement?Share a Content Correction