missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ BacLight™ Bacterial Membrane Potential Kit

Product Code. 10730964
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10730964

Brand: Molecular Probes™ B34950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

BacLight™ Bacterial Membrane Potential Kit

The BacLight™ Bacterial Membrane Potential Kit provides the fluorescent membrane-potential indicator dye, DiOC2(3), along with a proton ionophore (CCCP) and premixed buffer. DiOC2(3) at low concentrations exhibits green fluorescence in all bacterial cells, but it becomes more concentrated in healthy cells that are maintaining a membrane potential, causing the dye to self-associate and the fluorescence emission to shift to red. The red- and green-fluorescent bacterial populations are easily distinguished using a flow cytometer. CCCP is included in the kit for use as a control because it eradicates the proton gradient, eliminating bacterial membrane potential.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains DiOC2(3) (1.2 mL of 3 mM solution in DMSO), CCCP (300 μL of 500 μM solution in DMSO), and PBS (200 mL). Store in refrigerator (2–8°C) and protect from light.
Detection Method Fluorescence
For Use With (Application) Membrane Potential Assay
For Use With (Equipment) Flow Cytometer
Product Type Bacterial Membrane Potential Kit
Dye Type Membrane Potential Probes
Emission 482/497
Format Tube(s)
Product Line BacLight
Quantity 1 kit
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.