Learn More
Molecular Probes™ BacLight™ RedoxSensor™ Green Vitality Kit
BacLight™ RedoxSensor™ Green Vitality Kit
Brand: Molecular Probes™ B34954
Code : 50
Additional Details : Weight : 0.00525kg Transport : UN number : 3287 Chem class : 6.1 Pack group : III
Description
The RedoxSensor™ Green reagent included in the BacLight™ RedoxSensor™ Green Vitality Kit is an indicator of bacterial reductase activity. This reductase activity is, in turn, a reliable marker for changes in electron transport chain function and for changes in vitality that occurs following antibiotic treatment. RedoxSensor™ Green reagent penetrates both gram-positive and gram-negative bacteria. Following reduction, the RedoxSensor™ Green reagent will produce a stable green-fluorescent signal in 10 minutes that is compatible with formaldehyde fixation techniques.Specifications
Bacteria | |
Fluorescent | |
Green Vitality Kit | |
PI: 490⁄635, RedoxSensor™ green: 490⁄520 | |
Tube(s), Slide(s) | |
200 | |
DMSO (Dimethylsulfoxide) |
Contains 1 vial of RedoxSensor™ green (200 μL, 1 mM solution in DMSO), 1 vial of propidium iodide (300 μL, 20 mM solution in DMSO), 1 vial of sodium azide (1 mL of a 2 M solution in water), and 1 vial of CCCP (400 μL, 5mM in DMSO). | |
Fluorescence Microscope, Flow Cytometer | |
RedoxSensor™ green, PI | |
Solution | |
BacLight™, RedoxSensor™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.