Learn More
Cytiva E. coli BL21
Protease-deficient E. coli host strain for optimal expression of recombinant proteins
Brand: Cytiva 27-1542-01
Code : GE
Additional Details : Weight : 0.01500kg
Description
- F-, ompT, hsdS (rB-, mB-), gal, dcm
- Exemplary host well suited for use with the GST gene fusion system
- High-level expression of a variety of recombinant proteins
- Defective in E. coli strain B, and wild-type restriction systems to permit high efficiency transformation with unmodified plasmid DNA
- Verified by RAPD fingerprint analysis using Ready-To-Go™ RAPD analysis Kit
- Lacks the major Lon protease responsible for degradation of damaged or foreign proteins
Specifications
1 Vial |