missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ i-guard+ Planet Goggles
13.41 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Anti-fog, Chemical-resistant, Scratch-resistant
Description
The uvex i-range is our new flexible 3-in-1 safety eyewear line with the right product for many workplace hazards or environments. The uvex i-range planet variant also sets standards in terms of sustainability: we use recycled and bio-based materials for our innovative safety glasses as part of environmentally friendly and resource-saving production. The properties of our safety glasses (e.g. mechanical strength, UV protection, design and comfort) are not affected in any way. The uvex i-range planet is flexible in use and contributes effectively to environmental protection – true to our motto: protecting planet.
General features
- Versatile 3-in-1 protective eyewear range, which allows a flexible choice for many applications
- Large, flat lens design for an unrestricted field of vision
- Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
- Sustainably produced by the use of recycled and bio-based materials
- Metal-free
- Frame color: anthracite; headband: anthracite, turquoise
- PC lens: clear
Protection features
- Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
- Marking: W 166 34 B CE – 2C-1.2 W1 BKN CE
- UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
- Increased mechanical strength – B marking (120 m/s)
Comfort features
- X-tended sideshield offers additional side protection
- Soft, flexible face support that adapts to the wearer for optimal protection and comfort also protects against liquids and coarse dust (marking 3 and 4)
- Individually adjustable headband for a perfect fit and secure hold
Specifications
Lens Type | Chemical-resistant |
Lens Coating | Anti-fog, Scratch-resistant |
Certifications/Compliance (Europe) | EN 166, EN 170 |
Lens Tint | Clear |
Frame Color | Anthracite, Teal Green |
Headband Material | Recycled Materials, Textile |
Includes | Soft, Flexible Face Seal and Headband |
Lens Material | Polycarbonate |
UV Protection | UV400 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'