missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ i-guard+ Spectacles
13.96 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Lens Tint:
Clear
Gray
Description
- Member of the uvex i-range - versatile 3-in-1 safety eyewear range – allows eyewear to be adapted according to workplace hazards
- Large, flat lens delivers unrestricted field of vision
- X-tended sideshield offers additional protection from hazards
- Soft, flexible face seal adapts to the wearer providing both protection and comfort. It also protects against droplets or splashes of liquids and large dust particles (markings 3 and 4)
- Increased impact protection – markings BT
- Robust polycarbonate lens with uvex's proven supravision coating technology – permanent anti-fog and scratch resistant lens
- UV400 – 100 % protection against dangerous UVA and UVB radiation up to 400 nm
- Metal-free
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Color | Anthracite, Lime |
Certifications/Compliance (Europe) | EN 166, EN 170 |
Lens Tint | Gray |
Headband Material | Textile |
Lens Material | Polycarbonate |
UV Protection | UV400 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'