Learn More
Ambion™ mirVana™ miRNA Inhibitor
Product Code. 13416706
Click to view available options
Quantity:
20 nmol
250 nmol
5 nmol
Unit Size:
20nmol
250nmol
5nmol
Product Code. 13416706
Brand: Ambion™ 4464085
mirVana miRNA Inhibitors are single-stranded RNA molecules designed to inhibit endogenous microRNAs (miRNAs) and are ideal for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications.
mirVana miRNA Inhibitors are single-stranded RNA molecules designed to inhibit endogenous microRNAs (miRNAs) and are ideal for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications. Based on the latest version of miRbase, mirVana miRNA Inhibitors are available for a wide range of miRNAs, providing researchers with a powerful tool to investigate gene regulation mechanisms.
Key features include:
  • High specificity and potency—designed to closely resemble natural miRNAs, ensuring high specificity and potent gene regulation
  • Enhanced stability—chemical modifications enhance stability, making them suitable for long-term studies
  • Broad range of targets—available for a comprehensive list of miRNAs, allowing for extensive research possibilities
  • Optimized for delivery—compatible with various transfection reagents and delivery systems, ensuring efficient uptake by cells
miRNA inhibitors to increase gene expression
mirVana miRNA Inhibitors are molecules designed to specifically bind to and inhibit the activity of miRNAs. By inhibiting miRNAs, mirVana miRNA Inhibitors prevent them from binding to their target mRNAs, thereby allowing the expression of those genes to proceed. This can be useful in research to study gene function, understand disease mechanisms, and develop treatments that modulate gene expression.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C).
Description mirVana™ miRNA Inhibitor, 20nmol, Small, Chemically Modified Single-Stranded RNA Molecules, Designed to Specifically Bind to and Inhibit Endogenous miRNA, Molecules and Enable miRNA Functional Analysis by Down-Regulation of miRNA Activity, For Research Use Only
Product Type mirVana™ miRNA Inhibitor
Quantity 20 nmol