Learn More
Ambion™ mirVana™ miRNA Inhibitor
mirVana™ miRNA Inhibitor
Brand:  Ambion™ 4464084
200.33 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 13496696
  • £299.00 / 5nmol
Explore more special offers
Description
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in freezer (-5 to -30°C).
mirVana™ miRNA Inhibitors, 5nmol, Small, Chemically Modified Single-Stranded RNA, Inhibit Endogenous miRNA, Down-Regulation of miRNA Activity
5 nmol