missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ mirVana™ miRNA Inhibitor, let-7c positive control

miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.

£293.00 - £860.00

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer.
Control Type Positive Control
RNAi Type miRNA Inhibitor
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13315384
View Documents Promotion Details
Invitrogen™
4464080
5 nmol
£293.00
5nmol
Please to purchase this item. Need a web account? Register with us today!
13325384
View Documents Promotion Details
Invitrogen™
4464081
20 nmol
£860.00
20nmol
Please to purchase this item. Need a web account? Register with us today!
Description

Description

miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.

We have now released the second generation of miRNA inhibitors, mirVana™ miRNA Inhibitors, which:

  • Display better potency than lead competitor's product
  • Offer maximum and consistent effect in vitro at low miRNA inhibitor concentration
  • Target the most updated Sanger content
  • Can be used in vitro and in vivo, offering consistency throughout your entire research project
  • Are non-toxic

Positive and negative controls are also available for use along with mirVana™ miRNA Inhibitors in miRNA down-regulation experiments.

mirVana™ miRNA Inhibitor let-7c Positive Control

The mirVana™ miRNA Inhibitor let-7c Positive Control is intended for use as a positive control for mammalian cell culture experiments using mirVana™ miRNA Inhibitors. Endogenous let-7c miRNA has been shown to negatively regulate HMGA2 mRNA in cultured cells. HMGA2 is a ubiquitously expressed, nonhistone, chromatin protein that can modulate gene expression through changes in chromatin architecture. let-7c miRNA down-regulates levels of HMGA2 mRNA. When transfected into human and mouse cell lines, let-7c miRNA inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA. Thus, let-7c miRNA inhibitor activity can be monitored in human or mouse cells using real-time reverse transcription PCR (RT-PCR) to detect HMGA2 mRNA.

First Generation Inhibitor Controls

Our first-generation miRNA inhibitors (and controls), Ambion™ Anti-miR™ miRNA Inhibitors, have lower specificity and potency, plus a lower strand specificity compared to the new mirVana™ miRNA Inhibitors. These first-generation inhibitors are recommended for completing ongoing in vitro miRNA studies. For new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Inhibitors and their corresponding controls.

Specifications

Specifications

Store at or below -20°C. Do not store in a frost-free freezer.
miRNA Inhibitor
Unconjugated
Purified
Positive Control
miRNA & Non-Coding RNA Analysis
miRNA Inhibitor
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.