Learn More
Invitrogen™ mirVana™ miRNA Inhibitor, Negative Control #1
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
Brand: Invitrogen™ 4464076
193.63 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
We have now released the second generation of miRNA inhibitors, mirVana™ miRNA Inhibitors, which:
- Display better potency than lead competitor's product
- Offer maximum and consistent effect in vitro at low miRNA inhibitor concentration
- Target the most updated Sanger content
- Can be used in vitro and in vivo, offering consistency throughout your entire research project
- Are non-toxic
Positive and negative controls are also available for use along with mirVana™ miRNA Inhibitors in miRNA down-regulation experiments.
mirVana™ miRNA Inhibitor Negative Control #1
mirVana™ miRNA Inhibitor Negative Control #1 is intended for use as a negative control for experiments using mirVana™ miRNA Inhibitors. Transfect this negative control using the same methodology as for your positive control (such as let-7c Positive Control) and experimental mirVana™ miRNA Inhibitors. Use target expression from negative control-transfected samples as a baseline for evaluation of the effect of the control and experimental miRNA inhibitor on target gene expression.
First Generation Inhibitor Controls
Our first-generation miRNA inhibitors (and controls), Ambion™ Anti-miR™ miRNA Inhibitors, have lower specificity and potency, plus a lower strand specificity compared to the new mirVana™ miRNA Inhibitors. These first-generation inhibitors are recommended for completing ongoing in vitro miRNA studies. For new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Inhibitors and their corresponding controls.
Specifications
Store at or below -20°C. Do not store in a frost-free freezer. | |
miRNA Mimic | |
Unconjugated | |
miRNA Inhibitor | |
5 nmol |
Negative Control | |
miRNA & Non-Coding RNA Analysis | |
mirVana™ | |
Purified | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.