missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mirVana™ miRNA Inhibitor, Negative Control #1

Product Code. 15531511
Click to view available options
Quantity:
20 nmol
250 nmol
5 nmol
50 nmol
Unit Size:
20nmol
250nmol
50nmol
5nmol
Each
This item is not returnable. View return policy

Product Code. 15531511

Brand: Invitrogen™ 4464077

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

mirVana miRNA Inhibitor Negative Control No. 1 is a non-targeting control for miRNA inhibition experiments. It provides a baseline for evaluating effects on gene expression. Extensively tested in human cells, it ensures no impact on known miRNA functions.

mirVana miRNA Inhibitor Negative Control No. 1 is a non-targeting control designed for use in miRNA inhibition experiments. It provides a reliable baseline for evaluating the effects on target gene expression by ensuring minimal off-target effects. Extensively tested in human cell lines and tissues, this control is validated to have no impact on known miRNA functions, making it essential for accurate and consistent gene expression studies. Its properties include high reliability, superior specificity, and versatile use in various experimental setups.

Features of mirVana miRNA Inhibitor Negative Control No. 1 include:

  • Reliability—maximum and consistent effect in vitro at low concentration and low toxicity
  • Potency—better potency than lead competitor's product
  • Superior specificity—the most updated Sanger content
  • Versatile use—can be used in vitro and in vivo, offering you consistency throughout your entire research project

What are mirVana miRNA Inhibitors?

mirVana miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity. Positive and negative controls are also available for use along with mirVana miRNA inhibitors in miRNA down-regulation experiments.

Quality control

Each mirVana miRNA Inhibitor is synthesized and purified in state-of-the-art facilities to meet the highest quality standards. Our rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. We also assess each miRNA Inhibitor by gel electrophoresis to ensure a premium-quality miRNA inhibitor that is purified and ready for use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer.
Control Type Negative Control
RNAi Type miRNA Inhibitor
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
Product Line mirVana
Product Type miRNA Inhibitor
Purification Method Purified
Quantity 20 nmol
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.