missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ mirVana™ miRNA Mimic

Product Code. 13436696
Click to view available options
Quantity:
0.25 nmol
1 nmol
20 nmol
250 nmol
5 nmol
Unit Size:
20nmol
250nmol
5nmol
Each
This item is not returnable. View return policy

Product Code. 13436696

Brand: Ambion™ 4464067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity.

mirVana miRNA Mimics are designed to mimic endogenous microRNAs (miRNAs) and are excellent for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications.

mirVana miRNA Mimics are available for a wide range of miRNAs, providing researchers with a powerful tool to investigate gene regulation mechanisms.

Key features include:

  • High specificity and potency—designed to closely resemble natural miRNAs, helping ensure high specificity and potent gene regulation
  • Enhanced stability—chemical modifications enhance stability, making them suitable for long-term studies
  • Broad range of targets—available for a comprehensive list of miRNAs from miRBase v22, allowing for extensive research possibilities
  • Optimized for delivery—compatible with various transfection reagents and delivery systems, ensuring efficient uptake by cells
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Description mirVana™ Mimics and Inhibitors, 20nmol, Mimics, Which are More Specific than their Predecessors, miRNA Inhibitors, Which are More Potent than heir Predecessors, Enable miRNA Functional Analysis by Up-regulation of miRNA Activity
RNAi Type miRNA
Final Product Type mirVana™ Mimics and Inhibitors
Packaging Type 1 Each
Quantity 20 nmol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.