Learn More
Ambion™ mirVana™ miRNA Mimic
Description
mirVana miRNA Mimics are designed to mimic endogenous microRNAs (miRNAs) and are excellent for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications.
mirVana miRNA Mimics are available for a wide range of miRNAs, providing researchers with a powerful tool to investigate gene regulation mechanisms.
Key features include:
- High specificity and potency—designed to closely resemble natural miRNAs, helping ensure high specificity and potent gene regulation
- Enhanced stability—chemical modifications enhance stability, making them suitable for long-term studies
- Broad range of targets—available for a comprehensive list of miRNAs from miRBase v22, allowing for extensive research possibilities
- Optimized for delivery—compatible with various transfection reagents and delivery systems, ensuring efficient uptake by cells
Specifications
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Description | mirVana™ Mimics and Inhibitors, 250nmol, Small, Chemically Modified Double-stranded RNAs that Mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-regulation of miRNA Activity |
| Format | 96-well Plate, 24-well Plate, 6-well Plate |
| RNAi Type | miRNA |
| Final Product Type | mirVana™ Mimics and Inhibitors |
| Quantity | 250 nmol |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.