missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mirVana™ miRNA Mimic, Negative Control #1

Product Code. 15521124
Click to view available options
Quantity:
20 nmol
250 nmol
5 nmol
50 nmol
Packungsgröße:
20nmol
250nmol
50nmol
5nmol
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15521124

Marke: Invitrogen™ 4464061

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

mirVana miRNA Mimic Negative Control No. 1 is a non-targeting control for experiments with mirVana miRNA mimics. Providing a baseline for evaluating miRNA gene expression effects, this miRNA mimic negative control is validated to ensure no impact on known miRNA functions in human cells.

mirVana miRNA Mimic Negative Control No. 1 is a non-targeting control for experiments with mirVana miRNA mimics. Transfected using the same methodology as positive controls and experimental miRNA mimics, it provides a baseline for evaluating effects on target gene expression. Extensively tested in human cell lines and tissues, it ensures no impact on known miRNA functions, making it essential for accurate gene expression studies. Key properties include reliability, superior specificity, and versatile use in vitro and in vivo.

Features of mirVana miRNA Mimic Negative Control include:

  • Reliability—maximum and consistent effect in vitro at low concentration and low toxicity
  • Superior specificity—the most updated Sanger content with unique Star strand modification
  • Versatile use—can be used in vitro and in vivo, offering you consistency throughout your entire research project

What are mirVana miRNA mimics?

mirVana miRNA mimics are small, chemically modified double-stranded RNA molecules designed to mimic endogenous miRNA molecules and enable miRNA functional analysis by up-regulation of miRNA activity. Positive and negative controls are also available for use along with mirVana miRNA mimics in miRNA up-regulation experiments.

Quality control

Each mirVana miRNA Mimic is synthesized and purified in state-of-the-art facilities to meet the highest quality standards. Our rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. We also assess each annealed miRNA mimic by gel electrophoresis to ensure proper strand annealing. The result is a premium-quality miRNA mimic that is purified and ready for use.

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage Store at or below –20°C. Do not store in a frost-free freezer.
Control Type Negative Control
RNAi Type miRNA Mimic
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
Product Line mirVana
Product Type miRNA Mimic
Purification Method HPLC
Purity HPLC
Quantity 250 nmol
Shipping Condition Room Temperature
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt