missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ I/P™ Fixed-Speed Washdown Drive System

Masterflex™ I/P™ Fixed-Speed Washdown Drive System is IP55-rated for protection against dust and water

Brand:  Masterflex™ 07588-97

Product Code. 11896763

  • £8830.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flow rate up to 2.2 LPM per channel - flow rate depends on drive rpm, pump head, and tubing size
  • Drive comes equipped with either 3 or 4 I/P™ Easy-Load™ pump heads
  • Purge system or pump in either direction with combination on/off/reverse switch
  • IP55-rated housing is constructed of painted steel/aluminum and protects against dust and water
  • Fixed-speed drive features 150 rpm speed
  • Runs on 230 VAC, 50 Hz power
  • Pump system provides the option of three or four channels controlled through a single reversible fixed-speed motor
Specifications

Specifications

4 Included
1.9 LPM/Channel
Drive, Easy-Load™ Pump Heads (4), C-Flex™ ULTRA Tubing (10 ft.), Line Cord with European Plug (6 ft.)
Accepts I/P™ 26, 73, 82
230 VAC
Fixed Speed
150 rpm
Painted Steel (IP55-rated)
(76.91 x 28.47 x 26.11 cm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers