Learn More
Masterflex™ I/P™ Standard Pump Head Rotor Assembly
Masterflex™ I/P™ Standard Pump Head Rotor Assembly is constructed of durable Cold-Rolled Steel
Brand: Masterflex™ 07019-82
This item is not returnable.
View return policy
Description
- Get the most our of Masterflex™ I/P™ Standard Pump Heads with these replacement parts
- Choose replacement parts suitable for your I/P™ Standard pump head size
- Constructed of durable cold-rolled steel (CRS)
- Replacement parts to keep your pump head operating even longer
- Two end bell assemblies are required for a complete head assembly
Specifications
I/P™ Standard Pump Heads | |
Cold-Rolled Steel (CRS) | |
Short-Shaft |