Learn More
Masterflex™ I/P™ Tubing Loading Key
Masterflex™ I/P™ Tubing Loading Key ensures proper tubing alignment and tension
Brand:  Masterflex™ 07019-90
Product Code. 11715449
  • £20.00 / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with I/P™ Standard Pump Heads
  • Key is required to ensure proper alignment and tension
  • Constructed of durable stainless steel
Specifications
Specifications
Ensures proper tubing alignment and installation
I/P™ Standard Pump Heads
Stainless Steel
Replacement Key
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ I/P™ Tubing Loading Key > Stainless Steel