missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pichia Expression Kit, original kit

Product Code. 10768244
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10768244

Brand: Invitrogen™ K171001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The Original Pichia Expression Kit is designed for high-level expression of recombinant protein in the yeast Pichia pastoris.

The Original Pichia Expression Kit is designed for high-level expression of recombinant protein in the yeast Pichia pastoris. The vectors carry the HIS4 gene for selection of transformants on histidine-deficient medium. The Original Pichia Expression Kit provides all of the tools and reagents needed to express a gene of interest from the AOX1 promoter.

TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Ampicillin (AmpR)
Bacterial or Yeast Strain GS115, KM71
Content And Storage The Original Pichia Expression Kit includes 10 μg each of pHIL-D2 and pPIC3.5 vectors for intracellular expression and pHIL-S1 and pPIC9 vectors for secreted expression, Pichia pastoris strains GS115 (his4), KM71(arg4 his4 aox1::ARG4), control strain GS115 Albumin, control strain GS115 β-gal, media (YP and YNB), Spheroplast Transformation Kit, and 2 μg each 5' AOX1, 3' AOX1, and α-factor sequencing primers.

Store vectors and primers at -20°C. Store strains at room temperature.
All components are guaranteed stable for 6 months when properly stored., Store vectors and primers at -20°C. Store strains at room temperature. All components are guaranteed stable for 6 months
Cloning Method Restriction Enzyme⁄ MCS
Promoter AOX1
Cell Type Yeast Cells
Format Kit
Species P. pastoris
Vector pPIC
Selection Agent (Eukaryotic) Ampillicin
Expression Mechanism Cell-Based Expression
Expression System Yeast
For Use With (Application) Protein Expression
Product Type Expression Kit
Protein Tag Untagged
Quantity 1 Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.