missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Power SYBR™ Green RNA-to-CT™ 1-Step Kit
The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets.
£197.00 - £582.00
Specifications
Detection Method | SYBR |
---|---|
PCR Method | 1-step RT-qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10734227
|
Applied Biosystems™
4391178 |
40 Reactions |
£197.00
Each |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10229194
|
Applied Biosystems™
4389986 |
200 Reactions |
£582.00
200 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets.
- Get sensitivity and specificity in an easy-to-use one-step qRT-PCR reaction.
- Formulated to ensure maximum sensitivity and reliability
- Reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets
Includes:
- AmpliTaq™ Gold DNA Polymerase UP for hot start qPCR, minimizing non-specific product formation
- ArrayScript™ UP Reverse Transcriptase, an engineered RT that produces high yields of full-length cDNA
- An additive that reduces primer dimer formation, reducing non-specific products that can cause false positive signals
- RNase inhibitor, to prevent degradation of RNA templates
- A proprietary ROX™ dye that serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations for superb precision on Applied Biosystems™ real-time PCR instruments
Specifications
SYBR | |
AmpliTaq Gold DNA Polymerase | |
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System | |
ROX (Pre-mixed) | |
UP (Ultra Pure) | |
Standard | |
Gene Expression |
1-step RT-qPCR | |
Standard | |
SYBR Green | |
RNA-to-CT™, SYBR™ | |
ArrayScript™ UP | |
Wet Ice | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.