missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Cy™3-labeled Negative Control No. 1 siRNA

Product Code. 10026244
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10026244

Brand: Invitrogen™ AM4621

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Silencer™ Cy™3-labeled Negative Control No. 1 siRNA

Invitrogen™ A fluorescently labeled negative control siRNA for the easy monitoring of siRNA uptake by fluorescence microscopy. Purified, annealed, and ready to use, the 5 nmol aliquot is supplied in a stable, dried format.

  • Simplifies the monitoring of siRNA delivery
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use

    Visualize siRNA Uptake with Labeled siRNAs
    Fluorescent labeled siRNAs allow for the direct observation siRNA cellular uptake, distribution, and localization. Fluorescently labeled siRNAs are commonly used to monitor transfection efficiency during optimization of transfection conditions. In general, siRNAs are rapidly taken up by and distributed throughout cells in as little as 4 hours. Fluorescent labeling of siRNAs allows for rapid monitoring of siRNA delivery by fluorescence microscopy, as opposed to unlabeled siRNAs, whose delivery must be monitored by target gene knockdown. The Silencer™ Cy™3 Labeled Negative Control #1 siRNA sequence is identical to that of the Invitrogen Silencer™ Negative Control# 1 siRNA (SKU #AM4611), but with the fluorescein derivative Cy™3 (excitation max. = 547, emission max. = 563 nm), on the 5' end of the sense strand, making it highly fluorescent when excited by the appropriate wavelength of light. Silencer™ Cy™3 Labeled Negative Control #1 siRNA is suitable for monitoring siRNA uptake by chemical transfection or electroporation. Cy™3 labeling is recommended over FAM™ labeling when using electroporation because of its brighter fluorescence signal visible over a broader pH range. Electroporation produces a more diffuse siRNA distribution pattern, resulting in a less intense fluorescence signal than with chemical transfection.

    Accessory Products:
    Silencer™ FAM™ Labeled Negative Control #1 siRNA (SKU #AM4620) is also available. Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.
    Control Type Negative Control
    RNAi Type siRNA
    For Use With (Application) Transfection, RNAi
    Label or Dye Cy3
    Product Line Silencer, Ambion
    Product Type siRNA
    Purity HPLC
    Quantity 5 nmol
    Shipping Condition Dry Ice

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.