missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ FAM-labeled GAPDH siRNA (human, mouse, rat)

Product Code. 10702127
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10702127

Brand: Invitrogen™ AM4650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Silencer™ FAM-labeled GAPDH siRNA (human, mouse, rat)

Invitrogen™ fluorescently labeled control siRNA for the easy monitoring of siRNA uptake by fluorescence microscopy along with siRNA-induced knockdown. Purified, annealed, and ready to use, the 5 nmol aliquot is supplied in a stable, dried format.

  • Fluorescently labeled siRNAs simplify the monitoring of siRNA delivery
  • Ideal for correlating siRNA uptake with mRNA knockdown
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use

    Visualize siRNA Uptake with Labeled siRNAs
    Fluorescent labeling of siRNAs allows direct observation of their cellular uptake, distribution, and localization. Fluorescently labeled siRNAs are commonly used to monitor transfection efficiency during optimization of transfection conditions. In general, siRNAs are rapidly taken up by and distributed throughout cells in as little as 4 hours.

    Correlate siRNA Uptake with Gene Silencing
    Studies show that the silencing activity of Silencer™ FAM™ Labeled GAPDH siRNA is not affected by the fluorescent label. Therefore, these siRNAs are ideal for examining both siRNA uptake and siRNA-induced knockdown, and for correlating gene knockdown with transfection efficiency; this is important to know because different transfection reagents interact with siRNAs in different ways (i.e., some reagents may sequester the siRNA, resulting in poor gene knockdown). The Silencer™ FAM™ Labeled GAPDH siRNA sequence provides an easy way to monitor siRNA uptake by chemical transfection. The sequence is identical to that of Invitrogen Silencer™ GAPDH siRNA (SKU #AM4605), but with the fluorescein derivative FAM (5'-carboxyfluorescein; excitation max. = 494 nm, emission max. = 520 nm) on the 5' end of the sense strand, making it highly fluorescent when excited by the appropriate wavelength of light.

    Accessory Products:
    For monitoring siRNA delivery by electroporation, Silencer™ Cy™3 Labeled GAPDH siRNA (human) (SKU #AM4623) or Silencer™ Cy™3 Labeled GAPDH siRNA for human, mouse and rat (SKU #AM4649) is recommended. Cy™3 has a brighter fluorescence signal than FAM™, visible over a broader pH range. Electroporation produces a more diffuse siRNA distribution pattern resulting in a less intense fluorescence signal than with chemical transfection. Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.
    Control Type Positive Control
    RNAi Type siRNA
    Species Human, Mouse, Rat
    Sample Type siRNA
    Label or Dye FAM
    Product Line Silencer, Ambion
    Product Type siRNA
    Purity HPLC
    Quantity 5 nmol
    Shipping Condition Dry Ice
    Target GAPDH
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.