missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Firefly Luciferase (GL2 + GL3) siRNA

Silencer™ Firefly Luciferase (GL2 + GL3) siRNA

Brand:  Invitrogen™ AM4629

136.68 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10477874

  • £204.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Invitrogen™ Silencer™ Firefly Luciferase (GL2 + GL3) Positive Control siRNA is ideal for developing and optimizing siRNA experiments. The control is validated for use in human cell lines and optimized for mouse and rat. 5 nmol is provided, along with an additional tube of 2 nmol negative control siRNA.
  • Validated siRNA controls for optimizing siRNA experiments
  • Gene-specific control siRNAs provided with scrambled, negative controls
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Store at or below -20°C. Do not store in a frost-free freezer.
    siRNA
    Transfection, RNAi
    Silencer™, Ambion™
    HPLC
    Dry Ice
    Positive Control
    Firefly Luciferase
    Unconjugated
    siRNA
    5 nmol
    Luciferase
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.