missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Human Transcription Factor siRNA Library

Product Code. 15671852
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15671852

Brand: Invitrogen™ A30088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Transcription factors are fast becoming promising drug targets, as they bind to and regulate the transcription of genes.

Transcription factors are fast becoming promising drug targets, as they bind to and regulate the transcription of genes. The Silencer Human Transcription Factor siRNA Library targets genes expressing proteins that bind DNA in a sequence-specific manner to control the rate of transcription of genetic information from DNA to messenger RNA. The Silencer™ Human Transcription Factor siRNA Library contains a total of 5583 Ambion™ Silencer siRNAs targeting 1861 genes. The library was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC). Utilizing Ambion Silencer technology built on stringent bioinformatics to ensure high target specificity and siRNA potency, the Silencer Human Transcription Factor siRNA Library is an ideal solution for focused and cost-effective transcription factor RNAi screening.

  • Comprehensive—Expedite the discovery of transcription factor regulation of gene expression with pre-defined Silencer Human Transcription Factor siRNA Library
  • Reliable—Increase target screening accuracy with three individual siRNAs per gene target
  • Effective—Reduce off target impacts at low concentrations with proven Ambion Silencer siRNA design
  • Convenient—Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats

Why use the Silencer Human Transcription Factor siRNA Library?

Together with careful bioinformatics selection of gene targets, Silencer siRNAs are optimized for design and high quality synthesis with guaranteed silencing. The Silencer Human Transcription Factor siRNA Library contains our pre-designed and when available, validated siRNAs generated with maximum potency and specificity in mind so you can focus on data quality. All Silencer libraries are selected against the latest available databases and resources for up-to-date content. Full siRNA sequence information, as well as the exon targeted and each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID, plus results from any validation experiments performed using the included siRNAs is provided. Please contact us for the complete list of targets.

Silencer siRNA libraries allow you to screen efficiently and maximize data output

Ambion Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human Transcription Factor siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).

Flexibility to meet your workflow needs

Thermo Fisher Scientific offers comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.

TRUSTED_SUSTAINABILITY

Specifications

Description 5583 total siRNAs targeting 1861 genes plated onto 96-well plates (66 plates)
Content And Storage 5583 total siRNAs targeting 1861 genes plated onto 66 x 96-well plates

Store at or below -20°C in a non-frost-free freezer.
Format 96-well Plate
RNAi Type siRNA
Species Human
For Use With (Application) RNAi
Product Line Silencer
Product Type siRNA Library
Purity Standard
Quantity 1 Set
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.