missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Negative Control No. 1 siRNA

Product Code. p-7075572
Click to view available options
Quantity:
40 nmol
5 nmol
5 x 40 nmol
Unit Size:
5nmol
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10095994

Brand: Invitrogen™ AM4611

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Silencer Negative Control No. 1 siRNA is a bioinformatically designed, non-targeting siRNA crucial for siRNA experiments. Matching Silencer chemistry, it is an essential negative control siRNA for achieving clear, accurate gene silencing results.

Silencer Negative Control No. 1 siRNA is a bioinformatically designed, non-targeting siRNA used as a negative control siRNA. It distinguishes specific effects of siRNA-mediated gene silencing from non-specific procedural effects, ensuring minimal off-target effects and providing a reliable baseline for comparison. Validated through extensive testing, it has minimal impact on gene expression, cell proliferation, viability, and morphology, making it essential for accurate results in human, mouse, and rat cells.

Negative controls, such as non-targeting siRNAs, are critical for distinguishing the specific effects of siRNA-mediated gene silencing from non-specific effects caused by the experimental procedure itself. Silencer negative control No. 1 is designed to ensure minimal off-target effects, providing a robust baseline for comparing the results of siRNA-treated samples. This negative control siRNA shares the same chemical modifications as other Silencer siRNAs.

Features of Silencer Negative Control siRNAs include:

  • No significant sequence similarity to human mouse, or rat genes
  • Minimal effects on cell proliferation and viability
  • Matching Silencer™ siRNA chemistry

Validated through extensive testing, including microarray analysis and multi-parametric cell-based assays, Silencer Negative Control No. 1 has been proven to have minimal impact on gene expression, cell proliferation, viability, and morphology. This makes it an essential tool for researchers aiming to achieve accurate and consistent phenotypic data in human, mouse, and rat cells.

What are Silencer siRNAs?

Silencer siRNAs are designed for potency and specificity using a proprietary algorithm. These unmodified siRNAs are cost-effective siRNAs with guaranteed silencing.

Quality control

Each Silencer siRNA is synthesized and purified in state-of-the-art facilities to meet the highest quality standards. Our rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. We also assess each annealed siRNA by gel electrophoresis to ensure proper strand annealing. The result is a premium-quality siRNA that is purified and ready for use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer.
Control Type Negative Control
RNAi Type siRNA
Species Human, Mouse, Rat
For Use With (Application) Transfection, RNAi
Label or Dye Unconjugated
Product Line Silencer, Ambion
Product Type siRNA
Purity HPLC
Quantity 5 nmol
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.