missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Select Human Druggable Genome siRNA Extension Set V4

For the first time, you can obtain the benefits of Ambion™Silencer™ Select siRNAs in a convenient pre-plated, library format.

£21340.00 - £21840.00

Specifications

RNAi Type siRNA
Species Human
For Use With (Application) RNAi
Product Line Silencer™, Silencer™ Select, Ambion™
Product Type siRNA
View More Specs

Products 2
Product Code Brand Format Price Quantity & Availability  
Product Code Brand Format Price Quantity & Availability  
10330115
View Documents
Invitrogen™
4397921
96-well Plate
£21840.00
Each
Please to purchase this item. Need a web account? Register with us today!
10094924
View Documents
Invitrogen™
4397924
384-well Plate
£21340.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

siRNA
RNAi
siRNA
Multiple Targets
Human
Silencer™, Silencer™ Select, Ambion™
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Silencer™ Select Human Druggable Genome siRNA Extension Set V4

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.