missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Silencer™ Select Negative Control No. 1 siRNA
A non-targeting negative control siRNA with the same chemical modifications for enhanced efficacy as in other Silencer™ Select siRNAs.
£244.00 - £596.00
Specifications
Control Type | Negative Control |
---|---|
RNAi Type | siRNA |
For Use With (Application) | RNAi, Transfection |
Label or Dye | Unconjugated |
Product Line | Silencer™, Silencer™ Select, Ambion™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10025994
|
Invitrogen™
4390843 |
5 nmol |
£244.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10259444
|
Invitrogen™
4390844 |
40 nmol |
£596.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- A non-targeting negative control siRNA with the same chemical modifications for enhanced efficacy as in other Silencer™ Select siRNAs
- Validated siRNA controls for optimizing siRNA experiments
- GAPDH Positive Control functionally tested in several common cell lines
- Negative Controls functionally proven to have minimal effects on cell proliferation and viability
- Include Silencer™ Select modifications for enhanced specificity
- For use in human, mouse, and rat cells
- Upon receipt, store siRNAs in a non-frost-free freezer at or below -20°C
What are Silencer™ Select siRNAs?
- Silencer™Select siRNAs are our next-generation siRNAs
- These siRNAs incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemical modifications to yield siRNAs with unrivalled efficacy, potency, and specificity
- The result is fewer failed experiments due to poor silencing, and cleaner, more consistent phenotypic data
Silencer™ Select Negative Control siRNAs
- Negative control siRNAs are siRNAs with sequences that do not target any gene product
- They are essential for determining the effects of siRNA delivery and for providing a baseline to compare siRNA-treated samples
- These negative control siRNAs have been tested by microarray analysis and shown to have minimal effects on gene expression
- In addition, these two controls have been tested in multi-parametric, cell-based assays and are proven to have no significant effect on cell proliferation, viability, or morphology in the cell lines tested
Specifications
Negative Control | |
RNAi, Transfection | |
Silencer™, Silencer™ Select, Ambion™ | |
HPLC |
siRNA | |
Unconjugated | |
siRNA | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title