missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Silencer™ Select Negative Control No. 2 siRNA
Silencer™ Select Negative Control No. 2 siRNA
£245.00 - £600.00
Specifications
Content And Storage | The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. |
---|---|
Control Type | Negative Control |
RNAi Type | siRNA |
For Use With (Application) | RNAi, Transfection |
Label or Dye | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10239724
|
Invitrogen™
4390846 |
5 nmol |
£245.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321475
|
Invitrogen™
4390847 |
40 nmol |
£600.00
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ambion™ Silencer™ SelectNegative Control #2 siRNA is extensively validated and an ideal control for many aspects of an siRNA experiment.- Premium-quality siRNA, highly purified and ready to use
- Functionally proven to have minimal effects on cell proliferation and viability
- Include Silencer™ Select modifications for enhanced specificity
- For use in human, mouse, and rat cells
What are Silencer™ Select siRNAs?
Silencer™ Select siRNAs incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemical modifications, to yield siRNAs with unrivalled efficacy, potency, and specificity. The result is fewer failed experiments due to poor silencing, and cleaner, more consistent phenotypic data.
Silencer™ Select Negative Control #2 siRNA:
Negative control siRNAs—siRNAs with sequences that do not target any gene product—are essential for determining the effects of siRNA delivery and for providing a baseline to compare siRNA-treated samples. Silencer™ Select Negative Control #2 siRNA has been carefully designed and extensively tested, and has no significant sequence similarity to mouse, rat, or human gene sequences. This negative control siRNA has been tested by microarray analysis and shown to have minimal effects on gene expression. It has also been tested in multi-parametric, cell-based assays and shown to have no significant effect on cell proliferation, viability, or morphology in the cell lines tested.
Quality Control:
Silencer™ Select siRNAs are manufactured in a state-of-the-art facility under the highest quality standards. Rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. Each siRNA is also assessed by gel electrophoresis to confirm that the strands anneal properly.
Accessory Product: siPORT™ NeoFX™ Transfection Agent (SKU #AM4510 and #AM4511)
siPORT™ NeoFX™ Transfection Agent is a versatile, lipid-based transfection agent for the efficient transfection of siRNA into cultured cells. The protocol can be adapted to a wide range of cells and experimental designs, including high-throughput applications. Silencer™ Select siRNA controls have specific chemical modifications; they are designed to be used with Silencer™ Select siRNAs. If you are using other siRNA products, such as BLOCK-iT™ siRNAs, Ambion™ Silencer™ siRNAs, or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
Specifications
The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. | |
siRNA | |
Unconjugated | |
siRNA | |
Room Temperature |
Negative Control | |
RNAi, Transfection | |
Silencer™, Silencer™ Select, Ambion™ | |
HPLC |
For Research Use Only. Not for use in diagnostic procedures.