missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ siRNA Labeling Kit with Cy™3 dye

Product Code. 10299764
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10299764

Brand: Invitrogen™ AM1632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Silencer™ siRNA Labeling Kit with Cy™3 dye

For the labeling of siRNA with Cy™3. Includes sufficient reagents for labeling 65μg of siRNA. Invitrogen™ Labeled siRNA can be used to analyze siRNA subcellular localization, stability, and transfection efficiency. In addition, fluorescently labeled siRNA is particularly well-suited for use in double-label experiments (with a labeled antibody) to track cells that receive siRNA during transfection and to correlate transfection with down-regulation of the target protein. Research shows that labeling of siRNAs does not affect their biological function.

Accessory Products:
Also available is the Silencer™ FAM™ siRNA Labeling Kit (SKU# AM1634).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10X Labeling Buffer, Reconstitution Solution, 5M NaCl, 5X siRNA Annealing Buffer, GAPDH siRNA, and Cy™3 Labeling Reagent should be stored at –20°C. Nuclease-free water may be stored at any temperature.
Detection Method Fluorescence
Format Kit
Label or Dye Cyanine3
Final Product Type siRNA (Labeled)
Labeling Target siRNA
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line Silencer, Ambion
Product Type siRNA Labeling Kit
Quantity 1 kit
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.