missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq Buffers (10X)

Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.

Brand:  Thermo Scientific™ B55

11.99 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11859622

  • £18.44 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA polymerases (both recombinant and native). They are available in different compositions and also without detergents.

10X Taq Buffer without Detergent

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl

10X Taq Buffer with KCl and 15 mM MgCl2

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • 15 mM MgCl2
  • Detergent

10X Taq Buffer with (NH4)2SO4

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20

10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20
  • 20 mM MgCl2

10X Taq Buffer with KCl

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • Detergent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 10X Taq Buffer without Detergent, 4 x 1.25 mL

Store at -20°C.
Taq DNA Polymerase
4 x 1.25 mL
Wet or Dry Ice
10X
Liquid
Taq Buffer without Detergent
PCR
Standard PCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.