missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Taq DNA Polymerase, native, w/W1
Taq DNA Polymerase, native, w/W1
£71.00 - £238.00
Specifications
Content And Storage | –20°C |
---|---|
For Use With (Application) | Standard PCR |
Concentration | 5U/μL |
Overhang | 3'-A |
Source | Thermus aquaticus YT1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
10063172
|
Invitrogen™
18038034 |
100 Unit(s) | N/A | N/A | N/A | |||||
10594423
|
Invitrogen™
18038026 |
500 Unit(s) |
£238.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. The 1% solution of the detergent W-1 that is included may improve the thermostability of the enzyme when added at a final concentration of 0.05% (v/v).Taq DNA Polymerase consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5´→3´ DNA polymerase activity and a 5´→3´ exonuclease activity (see figure). With Taq DNA Polymerase, you get:
Applications
Amplification of DNA from complex genomic, viral, and plasmid templates, RT-PCR, sequencing ssDNA, and cycle sequencing
Source
Native enzyme is purified from Thermus aquaticus YT1. Unit definition
One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 min at 74°C.
Specifications
–20°C | |
5U/μL | |
Thermus aquaticus YT1 |
Standard PCR | |
3'-A |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Taq DNA Polymerase, native, w/W1