missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ i3300 Industrial Label Printer Kit UK with Wi-Fi and Workstation SFID Suite
The BradyPrinter i3300 Industrial Label Printer can print a wide range of sign and labels in monocolor, including labels to identify wires, laboratory samples, and facilities without any hassle.
Brand: Brady™ 241121
1271.20 GBP valid until 2024-12-31
Use promo code "21918" to get your promotional price.
This item is not returnable.
View return policy
Description
This mid-volume, i3300 Industrial Label Printer is ideal for identification throughout your facility, including everything from wires and panels to safety and facility labels.
- Wi-Fi connectivity allows you to get data more easily from a variety of sources
- Print smart technology allows printing on the first label- even sleeves and small labels
- Foolproof drop-in label rolls and ribbons snap into place in 20 seconds
- Prints on more than 825 parts and 70 material options for facility-wide identification
- Contact our Brady™ Technical Support Team to schedule a free phone appointment
- Includes UK power cord and Brady™ Workstation Safety & Facility Identification Suite
Specifications
Color LCD Touchscreen | |
9 in. | |
9.5 in. | |
12 in. | |
Not Autoclavable | |
i3300 Printer, UK Power Cord, Brady™ Workstation SFID Suite, USB Cable, Product CD, Cleaning Kit, Printed Quick Start Guide | |
195 MB | |
101.6 mm/s | |
Ethernet, USB, USB 2.0 | |
10°C to 40°C | |
5.2 kg | |
1 Year | |
i3300-C-UK-W-SFIDS |
Thermal Transfer | |
229 mm | |
241 mm | |
305 mm | |
Monochrome | |
Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Spanish, Swedish, Turkish | |
i3300 | |
300 dpi | |
Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016 | |
11.5 lb. | |
Barcode and Inventory Labeling, Hazard Identification, Laboratory Labeling | |
110/240 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction