missing translation for 'onlineSavingsMsg'
Learn More

Brady™ i3300 Industrial Label Printer Kit UK with Workstation PWID Suite

Product Code. 16386925
Click to view available options
:
i3300 Industrial Kit UK with PWID; Color Capability: Monochrome; Print Speed: 101.6 mm/s; Print Technology: Thermal Transfer; Resolution: 300 dpi; Weight (Metric): 5.2 kg
This item is not returnable. View return policy

Product Code. 16386925

Brand: Brady™ 241126

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The BradyPrinter i3300 Industrial Label Printer can print a wide range of sign and labels in monocolor, including labels to identify wires, laboratory samples, and facilities without any hassle.

This mid-volume, i3300 Industrial Label Printer is ideal for identification throughout your facility, including everything from wires and panels to safety and facility labels.

  • Print smart technology allows printing on the first label- even sleeves and small labels
  • Foolproof drop-in label rolls and ribbons snap into place in 20 seconds
  • Prints on more than 825 parts and 70 material options for facility-wide identification
  • Contact our Brady™ Technical Support Team to schedule a free phone appointment
  • Includes UK power cord and Brady™ Workstation Product & Wire Identification Suite

Specifications

Display Type Color LCD Touchscreen
Print Technology Thermal Transfer
Width (English) 9 in.
Width (Metric) 229 mm
Height (English) 9.5 in.
Height (Metric) 241 mm
Depth (English) 12 in.
Depth (Metric) 305 mm
Autoclavable Not Autoclavable
Color Capability Monochrome
Includes i3300 Printer, UK Power Cord, Brady™ Workstation PWID Suite, USB Cable, Product CD, Cleaning Kit, Printed Quick Start Guide
Languages Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Spanish, Swedish, Turkish
Memory 195 MB
Model i3300
Print Speed 101.6 mm/s
Resolution 300 dpi
Connector Types Ethernet, USB, USB 2.0
Operating System Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
Temperature Range (Metric) 10°C to 40°C
Weight (English) 11.5 lb.
Weight (Metric) 5.2 kg
For Use With (Application) Barcode and Inventory Labeling, Hazard Identification, Laboratory Labeling
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number i3300-C-UK-PWID
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.