missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IA-2/PTPRN Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-58897PEP
Additional Details : Weight : 0.00970kg
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PTPRN. The IA-2/PTPRN Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
5798 | |
>80% by SDS-PAGE and Coomassie blue staining | |
PBS and 1M Urea, pH 7.4. | |
PTPRN | |
100 μl | |
E. coli |
Human | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Antibody Competition | |
30 kDa | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54317. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |