Learn More
Ibalizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: N/A MA541841
Additional Details : Weight : 0.01000kg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Ibalizumab (also known asibalizumab-uiykand formerly known as TNX-355) is a monoclonal antibody that binds to CD4 receptors on the surface of CD4-positive cells, preventing HIV particle entry into the lymphocytes. It is an advanced and current antibody in development for the treatment of HIV/AIDS. It has been developed by Taimed biologics and Theratechnologies 8, 5.Specifications
Ibalizumab | |
Recombinant Monoclonal | |
Unconjugated | |
Hu5A8; TMB-355; TNX-355 | |
Human CD4 D3 domain | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG4 κ |